anti-goat-secondary-antibodies
Our anti-goat secondary antibodies are affinity-purified polyclonal and monoclonal antibodies with well-characterized specificity for goat immunoglobulin classes.  They are available in host species including mouse, rabbit, chicken, donkey and pig, and in the following formats:
  • Unlabeled antibodies
  • Enzyme conjugates (alkaline phosphatase (AP) or horseradish peroxidase (HRP))
  • Fluorescent conjugates (Alexa Fluor, DyLight, Qdot, Fluorescein (FITC), etc.)
  • Biotin
View all anti-goat secondaries
Our secondary antibodies are available as whole IgG or the F(ab')2 fragment. Each of these antibodies is affinity purified using its target antigen (the primary antibody class) coupled to an agarose support. This affinity purification eliminates nonspecific antibodies, resulting in high specificity and low background signal. Conjugated antibodies are affinity purified before labeling.
Selected secondary antibodies have been further purified by adsorbing the antigen-purified antibody to an affinity column containing immobilized serum proteins or IgG of selected species. This additional processing minimizes antibody cross-reactivity to the serum proteins of those selected species. These antibodies are designated as Cross Adsorbed or Highly Cross Adsorbed, based on the amount of processing which has occurred.
How to find a secondary antibody
Step 1: Use the tool at the top of the page to filter by target, application, target species
Step 2: View more filters to narrow results by host species, conjugate and other criteria
See all secondary antibodies
superclonal-sho
Superclonal Secondary Antibodies
Thermo Scientific Superclonal secondary antibodies represent a breakthrough in recombinant antibody technology designed to provide precise and accurate detection of mouse, rabbit and goat primary antibodies in a variety of applications. Each Superclonal secondary antibody is formulated and optimized to help achieve excellent results in ELISA, western blot, and cell imaging.
Resources
Product selection guides
Learning and support