Search
Sample Preparation and Plasma Protein Depletion
Single-step, single use protein depletion products for the removal of high-abundance human serum proteins from serum, plasma or CSF, before mass spec and/or gel analysis for biomarker discovery, validation, or diagnostics.
| ![]() | SDS-PAGE analysis of the CaptureSelect™ HumanPlasma14 depletion of abundant proteins from human plasma. M: P-6649 molecular marker LOAD: 100x diluted human plasma FT: plasma flow-through S: stripping of bound plasma fraction. The protein fractions were stained with Sypro ruby dye. |
Products:
| Product | Target |
| CaptureSelect™ HumanPlasma14 | Depletes the top 14 most abundant proteins from human blood plasma: albumin, IgG, IgM, IgA, IgE, IgD and free light chains, transferrin, fibrinogen, α-1 antitrypsin, apolipoprotein A1, α-2 macroglobulin, α-1 acid glycoprotein, and haptoglobin; enables >99% depletion of albumin and other high-abundance proteins, with no nonspecific binding and the highest reported loading capacity |
| CaptureSelect™ HumanPlasma Albumin | Depletes albumin from human plasma or serum |
| CaptureSelect™ HumanPlasma Ig/Albumin | Depletes human plasma or serum of albumin and all Igs, including IgG, IgA, IgM, IgD, IgE, and free light chains |
| CaptureSelect™ Multi Species Albumin | Depletes albumin from plasma or serum derived from a number of different species (e.g., mouse, rat, rabbit, goat, bovine, guinea pig, human, and horse) |
Plasma Sample Prep Literature:
For more information on CaptureSelect™ products and services please contact us.