Publication-quality imaging in seconds
EVOS cell imaging stations were designed to eliminate the complexities of microscopy without compromising performance. These units combine all aspects of an inverted microscope workstation into a single, compact device that turns on with one switch and can be mastered in minutes. Whether you’re capturing images for publication, teaching, or research, these systems are designed to be easier, smarter, and faster.
No matter your application or budget, there’s an EVOS cell imaging system that brings simplicity right to your bench.
Request more information  Request a demo
Which cell imaging system is right for you?
  M7000 M5000 FLoid XL Core
  Cat. No. AMF7000 Cat. No. AMF5000 Cat. No. 4471136 Cat. No. AMEX1000
  Fluorescence solutions Bright-field solutions
Hardware attributes
Simple installation Yes Yes Yes Yes
Installation and training Service team User User User
Motorized encoded X/Y scanning stage Yes No    
Manual mechanical stage   Yes Yes (glide stage) No
Mechanical or fixed stage option       Yes
Objective turret positions 5 5 1 4
Objective range 1.25–100x 1.25–100x 20x 1.25–100x
Fluorescence channels 4 4 3 N/A
Fluorescence LED light cubes Yes Yes LED lights N/A
Monochrome or color camera option (built-in, not optional) Both Mono with LED-based RGB illumination scheme Mono Color
Epifluorescence images Yes Yes Yes No
Transmitted light images Yes Yes Yes Yes
Color images Yes Yes No Yes
Benchtop system Yes Yes Yes Yes
Suitable for use in tissue culture hood No Yes Yes Yes
Darkroom needed No No No No
Associated printer     Yes  
Onstage incubator for time-lapse imaging Optional Optional    
Time-lapse imaging Multichannel Multichannel    
Autofocus Yes Yes    
Z-stack capability Yes Yes    
Automated multiwell plate screening Yes      
Thermo Fisher Cloud connection   Yes    
USB ports Yes Yes Yes Yes
DVI ports   Yes    
Computer Stand-alone PC Embedded Embedded Embedded
Software attributes
Celleste analysis software Optional Off-system optional Off-system optional Off-system optional
Embedded analysis   Yes    
Intuitive onboard software Yes Yes Yes Yes
Networking capability Yes Yes Yes  
Integrated reagent selection guide     Yes