F-actin and Oxphos Complex V inhibitor protein in muntjac skin fibroblasts
At a glance . . .
Summary
Filamentous actin in muntjac skin fibroblasts was visualized using green-fluorescent Alexa Fluor® 488 phalloidin. Mitochondria were labeled with an anti-OxPhos Complex V inhibitor protein antibody and visualized using red-orange fluorescent Alexa Fluor® 555 goat anti-mouse IgG (pseudocolored magenta). Nuclei were stained with red fluorescent TO-PRO®-3.
Methods and Materials
Cells
Muntjac skin fibroblasts
Materials Needed
Protocol
Sample Preparation
(All steps are carried out at room temperature unless otherwise noted)
Sample Preparation
Staining
Mounting Reagent Preparation and Sample Processing
To view the samples immediately, secure the coverslip at the corners using nail polish or hot wax to prevent the coverslip from moving. Leave the edges clear to allow the preparation to cure.
Note: The antifade properties of ProLong® Gold antifade reagent improve slightly the longer it remains in contact with the specimen. To further reduce photobleaching, minimize the exposure of fluorescently labeled specimens to light by using neutral density filters, and expose samples only when observing or recording a signal. Optimize image capture by using a minimum of optics, high-numerical aperture objectives, relatively low magnification, high-quality optical filters, and high-speed film or high-efficiency detectors.
To further reduce photobleaching, minimize the exposure of fluorescently labeled specimens to light by using neutral density filters and expose samples only when observing or recording a signal. Optimize image capture by using a minimum of optics, high-numerical aperture objectives, relatively low magnification, high-quality optical filters, and high-speed film or high-efficiency detectors.
Muntjac skin fibroblasts
Materials Needed
- Phosphate-buffered saline (PBS) (GIBCO®)
- 10X Phosphate-buffered saline (PBS)(GIBCO®)
- Triton X-100 (Sigma)
- Methanol-free 16% formaldehyde solution (Polysciences)
- BlockAid™ Blocking Solution (Molecular Probes®)
- Bovine serum albumin (BSA; Sigma)
- Alexa Fluor® 488 phalloidin (Molecular Probes®)
- Anti–OxPhos Complex V inhibitor protein, monoclonal 5E2 (Molecular Probes®)
- Alexa Fluor® 555 goat anti–mouse IgG (Molecular Probes®)
- TO-PRO®-3 (Molecular Probes®)
- ProLong® Gold Antifade Reagent (Molecular Probes®)
Protocol
Sample Preparation
(All steps are carried out at room temperature unless otherwise noted)
Sample Preparation
- Remove media from live cells, wash in PBS, and fix cells for 15 minutes in 3.7% formaldehyde solution at 37°C.
- Wash cells 3 x 5 minutes in PBS (1X).
- Permeabilize cells by treating for 10 minutes with PBT (0.1% Triton X-100 in PBS (1X)).
- Wash cells 3 x 5 minutes in PBS.
Staining
- Block cells for 1 hour in antibody blocking buffer (BlockAid Blocking Solution).
- Incubate cells with 5 µg/ml of anti–OxPhos Complex V inhibitor protein primary antibody in BlockAid Solution for 1 hour.
- Wash cells 3 x 10 minutes in PBT.
- Incubate cells with 5 µg/ml Alexa Fluor 555 goat anti-mouse IgG in 1% BSA/PBT for 1 hour.
- Wash 3 x 10 minutes in PBS.
- Incubate cells for 20 minutes with Alexa Fluor 488 phalloidin, diluted 1:200 from stock solution.
- Wash 3 x 5 minutes in PBS.
- Label cells with 1 µM TO-PRO®-3 dye in PBS for 15 minutes.
- Wash 3 x 10 minutes in PBS.
- Mount the specimen in ProLong Gold antifade reagent.
Mounting Reagent Preparation and Sample Processing
- Remove the ProLong Gold antifade reagent from the freezer and allow the vial to equilibrate to room temperature. Using an external heat source to warm the vial is not recommended, as this may decrease the long-term stability of the product.
- Remove any excess liquid from the specimen and apply 1 or 2 drops (depending on the surface area of your sample) of the antifade reagent to the specimen. Cover slide-mounted specimens with a coverslip; for specimens mounted on coverslips, place a drop of antifade reagent onto a clean slide and carefully lower the coverslip onto the antifade reagent to avoid trapping any air bubbles.
- Allow the mounted sample to cure on a flat surface in the dark. Curing time may vary from a couple of hours to overnight, depending on the thickness of the sample and the relative humidity of the surrounding air. For long-term storage, seal the coverslip to the slide after curing to prevent excessive shrinkage of the mounting medium, which can result in sample distortion. After sealing, store the slide upright in a covered slide box at <=-20°C. Desiccant may be added to the box to ensure that the slide remains dry.
To view the samples immediately, secure the coverslip at the corners using nail polish or hot wax to prevent the coverslip from moving. Leave the edges clear to allow the preparation to cure.
Note: The antifade properties of ProLong® Gold antifade reagent improve slightly the longer it remains in contact with the specimen. To further reduce photobleaching, minimize the exposure of fluorescently labeled specimens to light by using neutral density filters, and expose samples only when observing or recording a signal. Optimize image capture by using a minimum of optics, high-numerical aperture objectives, relatively low magnification, high-quality optical filters, and high-speed film or high-efficiency detectors.
To further reduce photobleaching, minimize the exposure of fluorescently labeled specimens to light by using neutral density filters and expose samples only when observing or recording a signal. Optimize image capture by using a minimum of optics, high-numerical aperture objectives, relatively low magnification, high-quality optical filters, and high-speed film or high-efficiency detectors.
Table 1 - Instruments
Microscope | Objective Used |
---|---|
Nikon Eclipse 800 with MicroMax 1300 YHS 12-bit digital camera | 60x 1.40 NA oil |
Filters Used | ||
---|---|---|
Alexa Fluor® 488 | Alexa Fluor® 555 dye | TO-PRO®-3 dye |
Excitation: 475AF40 | Excitation: 535AF18 | Excitation: 630AF25 |
Dichroic: 505DRLP | Dichroic: 570DRLP | Dichroic: 650DRLP |
Emission: 535AF45 | Emission: 590AF18 | Emission: 695AF28 |
For Research Use Only. Not for use in diagnostic procedures.
Download Wallpapers
Related Links
More In-Depth Images
Cytokeratin and nuclei in mouse trophoblast giant cells
α - catenin in HeLa human cervical cancer cells
Golgi, f-actin, nuclei, and mitochondrial OxPhos Complex V Inhibitor Protein in NIH 3T3 cells
Goblet and Paneth cells, f-actin, and nuclei in mouse intestine
Myelin, Nissl bodies, and nuclei in mouse brain
α - catenin in HeLa human cervical cancer cells
Golgi, f-actin, nuclei, and mitochondrial OxPhos Complex V Inhibitor Protein in NIH 3T3 cells
Goblet and Paneth cells, f-actin, and nuclei in mouse intestine
Myelin, Nissl bodies, and nuclei in mouse brain