ArrayScan™ Liquid Handling Module
![]() |
Capture cellular phenotypes and responses of rapidly developing biological processes. Liquid handling and on-board pipetting enables the direct measurement of quickly developing biologies, kinetic studies of biology and combination target studies resulting in comprehensive pharmacological data down to the individual cell level. This module is designed to work with the Thermo Scientific™ ArrayScan™ Live Cell Module, expanding the breadth of cell processes that can be monitored. Full function aspirate, dispense, and mixing are available from 4 deck locations. The module is capable of single tip aspirate and dispense at any location, creating a highly flexible instrument. Compatible with 96- and 384-well plates, the Liquid Handling Module utilizes disposable tips with a dispense volume range of 3–200 μL. |
Features
- Compatible with 96- and 384-well plates, the module utilizes disposable tips with a dispense volume range of 3–200 μL
- Full function liquid handling from any plate or trough
- Industry standard arm with single pipette tip driven by syringe pump (e.g., aspirate, dispense)
- Compatible with all ArrayScan™ Modules
- Interchangeable with the ArrayScan™ Brightfield Module
- Dynamic access to compound and assay plate
- One-to-many and many-to-one transfer scenarios are supported
Specifications
Ambient temperature operating range | Permissible range during operation: 18° to 25°C |
---|---|
Compatibility | ArrayScan™ HCA Readers |
Deck positions | 3 plate and 1 quarter trough |
Description | Automated, single-tip, liquid handling robot module for the ArrayScan™ instrument using disposable tips (standard and liquid level sensing) to both aspirate and dispense from multiple deck and live cell chamber locations |
Dimensions (of ArrayScan™ HCA Reader including module) |
Height: 109.22 cm (43 in) Width: 87.63 cm (34.5 in) Depth: 74 cm (29 in) |
Microplate type | SBS conforming plates, up to 384-well; 6–22 mm plate height |
Operating humidity range | 75% at 30°C |
Relative humidity | Permissible relative humidity during operation (non-condensing): less than 75% at 30°C |
Software | Compatible with Thermo Scientific™ HCS Studio™ Cell Analysis Software |
Volume dispense | 3–200 µL ( ± 5% in 1 µL increments) |
Wattage | 150 W (+500 W from ArrayScan™ Reader) |
Weight | 59 kg (130 lb) |
Ordering Information
Product | Quantity | Catalog # | |
---|---|---|---|
ArrayScan™ Liquid Handling Module | 1 each | N010117A | Request a quote |
For Research Use Only. Not for use in diagnostic procedures.