ArrayScan™ Live Cell Module
![]() |
The ArrayScan™ Live Cell Module provides complete quantitative, kinetic, live-cell imaging and analysis for Thermo Scientific™ ArrayScan™ HCA Readers. Study the dynamics of intracellular molecular interactions and cell motion in an environment similar to that of a tissue culture incubator. The ArrayScan™ Live Cell Module offers full control of temperature, carbon dioxide, and humidity levels to maintain cell health and viability, helping to ensure more reliable results. |
Features
- Full environmental control—complete environmental control (CO2, humidity, and temperature) with full reporting of CO2 and temperature over short or extended periods of time
- Kinetic capabilities—cell tracking with powerful kinetic analysis software provides numerous motility and kinetic calculations at the cell and well levels
- Focus on cell health—Zeiss™ Definite Focus is a hardware-based autofocus system that maintains focus and experimental reliability while supporting long-term imaging of live cells; phototoxicity is reduced by substituting software focusing with less harmful near-IR LED technology
Specifications
Autofocus system | Hardware-based Zeiss™ Definite Focus autofocus system |
---|---|
Compatibility |
Thermo Scientific™ ArrayScan™ HCA Readers
|
CO2 mixed gas | Software controlled; 0 to 10% saturation with less than 0.1% variation measured across a plate; CO2 saturation recorded and reported through software at each time point |
External control | Module: D x W x H: 56 x 33 x 36 cm control unit for temperature and CO2 control |
Relative humidity | Maintains greater than 90% relative humidity |
Software | Thermo Scientific™ HCS Studio™ Cell Analysis Software |
Temperature control | Software controlled—ambient to 45°C; temperature variation is less than 0.5°C over 48 hours; temperature recorded and reported through the software at each time point |
Ordering Information
Catalog # | Product | Quantity | |
---|---|---|---|
N010114A | Thermo Scientific™ ArrayScan™ Live Cell Module | 1 each | Request a quote |
For Research Use Only. Not for use in diagnostic procedures.