Distilled Water
The water used in liquid cell culture products is distilled water meeting all USP monograph test requirements for Water For Injection. The system includes water pretreatment followed by distillation. Continuous quality assurance testing for microbial bioburden, endotoxin, and Water For Injection USP monograph requirements ensures that all specifications are met.
Membrane filtered and endotoxin screened.
Membrane filtered and endotoxin screened.