Making a Gene or Genetic Element
Making a gene or genetic elements can be done through cloning with TOPO® technology, constructing the gene with the GeneArt® Seamless Cloning and Assembly Kit or purchasing an optimized GeneArt® gene. TOPO® cloning is the ideal solution for the researcher who needs to clone genes with a high cloning efficiency. The GeneArt® seamless cloning allows a researcher to clone up to 4 fragments directly into any vector of their choice with high efficiency in a 30 minute bench-top cloning reaction. GeneArt® Optimized Genes are the most complete solution for customers, providing what they need with a minimum of hand-on time.
GeneArt® Seamless Cloning and Assembly Kits work great together with GeneArt® Strings™ DNA Fragments. This new service offers custom-made DNA fragments up to 1,000 bp ready for cloning and assembly and delivered in only 5-7 business days.
| Quick & simple cloning when constructing a gene | The highest efficiency, direct cloning technology available today | Custom optimized gene, 100% accurate, no limitation in sequence design | |
|---|---|---|---|
| TOPO® Cloning Learn more > |
GeneArt® Seamless Cloning and Assembly Kit & GeneArt® Strings™ DNA Fragments Learn more > |
GeneArt® Optimized Genes Learn more > |
|
| Key Features | Fastest cloning method with high efficiency into a cloning vector | Can clone up to 4 fragments into any vector without additional sequence | Full service that delivers validated Genes with improved expression yields, no limitation in sequence design |
| Time spent cloning | 5 minutes | 30 minutes | No hands-on time required at all |
| Maximum Number of Fragments | 1 | 4 | Service can deliver length of 20kb and more |
| Cloning Efficiency | 95% | 90% (single fragment); 75% (4 fragments) |
Service can deliver length of >20kb |
| Time to Clone Multiple Fragments | N/A | 1–3 days | 6–15 days to receive clones ready for use |
| Improvement of expression yields | N/A | N/A | Up to 100% |
| Webtool for oligo and construct design | N/A | Free! | Free! |
| Order Now | Order Now | Configure your project |
For Research Use Only. Not for use in diagnostic procedures.