More intuitive & faster sequence analysis suite; also runs on Macintosh | Integrated, multifunctional sequence analysis suite | ||
---|---|---|---|
|
|
||
PC Compatible version | ![]() |
![]() |
|
Macintosh compatible version | ![]() |
||
Automated software update delivery | ![]() |
||
Sequencing data compatibility | CE & next-generation sequencing | CE sequencing | |
Sequence Analysis using: | Blast, Entrez searching & Align X | Blast, Entrez searching & Align X | |
Sequence-based searches | ![]() |
![]() |
|
Integrated primer design & product ordering (e.g., TaqMan® Assays) | ![]() |
![]() |
|
Bioinformatics algorithms | EMBOSS standard-based algorithms | Proprietary | |
Protein Analysis Tools (predictive motiff mapping, 3D structure visualization) | ![]() |
![]() |
|
Store & manage data collections |
• DNA & Protein sequences
• Plasmid maps • Genomic DNA • Public & proprietary sequences & constructs |
• DNA & Protein sequences
• Plasmid maps • Genomic DNA • Public & proprietary sequences & constructs |
|
Sequence assembly |
• Primer design
• PCR & resequencing • Gel simulations • New subclone vector sequence & maps Cloning Tools: • Restriction enzyme & ligation (Clone2Seq) • TOPO® Cloning • Gateway® Cloning • GeneArt® Seamless & High-Order Assembly |
• Primer design
• PCR & resequencing • Gel simulations • New subclone vector sequence & maps Cloning Tools: • Restriction enzyme & ligation (Clone2Seq) • TOPO® Cloning • Gateway® & Multi-site Gateway® • GeneArt® Seamless & High-Order Assembly |
|
Chromosomal mapping & annotation using GenomBench™ |
• Visualization & analysis of mega-base sized genomic DNA fragments
• cDNA-to-Gene alignment • Intro-Exon boundary mapping |
• Visualization & analysis of mega-base sized genomic DNA fragments
• cDNA-to-Gene alignment • Intro-Exon boundary mapping |
|
Get a quote | Get a quote |