Plasmid DNA Midiprep Kits
Starting with high-quality plasmid DNA is critical to your workflow.
The table below will help you choose the right plasmid DNA midiprep kit.
Which plasmid DNA midiprep kit is right for you?
  Order now Order now Order now
  Quick and cost effective Eliminates the use of chaotropic salts Rapid purification with integrated lysate clarification filter
  GeneJET Plasmid Midiprep Kit PureLink™ HiPure Plasmid Midiprep PureLink™ HiPure Filter Plasmid Midiprep
Purity grade Molecular Transfection Transfection
Total protocol time 60 min 120 min 90 min
Yield Up to 200 µg Up to 200 µg Up to 200 µg
Technology Silica membrane Anion-exchange gravity-flow column Anion-exchange gravity-flow column
Clarification Filter No No Yes, save 30 minutes
Prep size 25, 100 preps 25, 50 preps 25, 50 preps
Resources
Nucleic Acid Purification and Analysis Support Center
Find tips, troubleshooting help, and resources for your nucleic acid purification & analysis applications.
DNA Purification Brochure
RNA Isolation & Purification Brochure