pSilencer™ 3.1-H1 hygro
AM5766: pSilencer™ 3.1-H1 hygro
Features:
Vector Size: 4552 bp
Promoter: Human H1
Ampicillin resistance gene
Hygromycin resistance gene (SV40 early promoter, SV40 early polyadenylation signal)
ColE1 origin of replication
Vector Map and Related Information
Features:
Vector Size: 4552 bp
Promoter: Human H1
Ampicillin resistance gene
Hygromycin resistance gene (SV40 early promoter, SV40 early polyadenylation signal)
ColE1 origin of replication
Vector Map and Related Information
Note: This is the sequence of the negative control vector provided with the kit. It contains a negative control siRNA template (66 base pairs) between the BamH I and Hind III sites.
Sequence:
For Research Use Only. Not for use in diagnostic procedures.