pSilencer™ adeno 1.0-CMV System
AM5790: pSilencer™ adeno 1.0-CMV System
PROPRIETARY -- DO NOT DISCLOSE
Description:
Shuttle Vector Sequence
We recommend the following primers for sequencing:
5' Sequencing Primer: 5'-ggatttccaagtctccac-3'
3' Sequencing Primer: 5'-agcaccttccagatctgc-3'
Features:
Vector Size: 5895 bp
Promoter: CMV
Ampicillin resistance gene
PolyA terminator
ColE1 origin of replication
Vector Map and Related Information
Sequence:
Backbone Sequence:
For Research Use Only. Not for use in diagnostic procedures.