E-Gel Precast Agarose Gels
E-Gel® agarose gels are self-contained, pre-cast agarose gels. Each E-Gel agarose gel is ready for use with agarose, electrodes, and DNA stain (ethidium bromide or SYBR Safe™) packaged inside a dry, disposable, UV-transparent cassette. There are no gels to pour, buffers to make, staining or destaining steps to perform, or gel boxes to assemble. Just load your samples and run. E-Gel agarose gels offer excellent resolution and clarity in as little as 12 minutes and are ideal for analyzing PCR products, restriction digests, and plasmid preparations.
E-Gel agarose gels are 12-well, single comb gels that are available in a variety of formats: with SYBR Safe DNA Gel Stain or ethidium bromide, with agarose percentages suitable for general purpose (0.8%, 1.2%, and 2%) or high resolution (4%) separations.
E-Gel Single Comb and Double Comb Gels
These gels are general-use, ready-to-use gels with agarose, electrodes, and nucleic acid stain packaged inside a dry, disposable, UV-transparent cassette. They are offered with a single row of 12 sample wells, or with two rows, each containing 8 sample wells and one marker well, and are spaced for compatibility with 8-channel pipettors for easy multi-well loading.
Order E-Gel Single Comb and Double Comb Gels
Product | Stain | #Sample wells |
Run time | Resolution | Cat. No. |
---|---|---|---|---|---|
E-Gel® 0.8% General Purpose Agarose 18-Pak | Ethidium bromide | 12 sample | 30 min | 800 bp–10 kb | G501808 |
E-Gel® 1.2% General Purpose Agarose 18-Pak | Ethidium bromide | 12 sample | 30 min | 100 bp–5 kb | G501801 |
E-Gel® 2% General Purpose Agarose 18-Pak | Ethidium bromide | 12 sample | 30 min | 100 bp–2 kb | G501802 |
E-Gel® 4% High-Resolution Agarose 18-Pak | Ethidium bromide | 12 sample | 30 min | 20 bp–500 bp | G501804 |
E-Gel® 0.8% double comb 18-Pak | Ethidium bromide | 16 sample & 2 marker | 15 min | 1 kb–10 kb | G601808 |
E-Gel® 2% double comb 18-Pak | Ethidium bromide | 16 sample & 2 marker | 15 min | 100 bp–2 kb | G601802 |
E-Gel® 0.8% Starter Pak | Ethidium bromide | 12 sample | 30 min | 800 bp–10 kb | G600008 |
E-Gel® 1.2% Starter Pak | Ethidium bromide | 12 sample | 30 min | 100 bp–5 kb | G600001 |
E-Gel® 2% Starter Pak | Ethidium bromide | 12 sample | 30 min | 100 bp–2 kb | G600002 |
G551801 Clear E-Gel® 1.2% 18-Pak* | None | 12 sample | 30 min | 100 bp–5 kb | G551801 |
E-Gel with SYBR Safe Gels
E-Gel with SYBR Safe Gels contain SYBR Safe DNA stain instead of ethidium bromide. The detection sensitivity of E-Gel gels with SYBR Safe stain is similar to that of gels containing ethidium bromide. Use E-Gel gels with SYBR Safe stain to:
- Minimize hazardous waste
- Eliminate the use of a strong mutagen (ethidium bromide) and reduce UV exposure
- Use blue-light transillumination to dramatically reduce DNA damage
Order E-Gel with SYBR Safe Gels
Product | Stain | #Sample wells | Run time | Resolution | Cat. No. |
---|---|---|---|---|---|
E-Gel® 1.2% with SYBR Safe™, 18-Pak | SYBR® Safe | 12 sample | 30 min | 100 bp–5 kb | G521801 |
E-Gel® 2% with SYBR Safe™ 18-Pak | SYBR® Safe | 12 sample | 30 min | 100 bp–2 kb | G521802 |
E-Gel® 1.2% with SYBR Safe™ Starter Kit | SYBR® Safe | 12 sample | 30 min | 100 bp–5 kb | G620601 |
E-Gel® 2% with SYBR Safe™, Starter Kit | SYBR® Safe | 12 sample | 30 min | 100 bp–2 kb | G620602 |
For Research Use Only. Not for use in diagnostic procedures.