Multidrop Combi+, Multidrop Combi SMART+ and Multidrop Combi nL
The Multidrop Combi+ and Multidrop Combi SMART+ are advanced reagent dispensers designed for high-throughput laboratories. Offering precise, flexible dispensing for a wide range of applications and ensuring accuracy and reliability for enhanced workflow efficiency. The Multidrop Combi nL Reagent Dispenser is a highly reliable and easy-to-use solution designed for low-volume reagent dispensing in pharmaceutical and biotechnology laboratories. It offers precise dispensing, ensuring high-quality assay data and improved laboratory efficiency.
Features
Featured videos
Get a look inside the Multidrop Combi+ and Combi SMART+
Note: This tool is best viewed in Chrome, Firefox or Safari Browsers
Applications
  • Assay development
  • Cell-based assays
  • ELISA
  • Genomic and Proteomic Research
  • Compound Storage
  • Primary and Secondary Screening
FILLit Software
Thermo Scientific FILLit Software is a cutting-edge solution designed to enhance the control and efficiency of the Multidrop Combi+ and Multidrop Combi SMART reagent dispensers. This intuitive PC software simplifies the setup and management of instruments, providing a streamlined experience that meets stringent quality control (QC) requirements. With its user-friendly interface and robust functionality, FILLit Software empowers laboratories to optimize their workflows, ensuring precision and reliability in liquid handling applications.
Advanced protocol development
  • Effortlessly develop, transfer, and store dispensing protocols, ensuring consistency and accuracy across experiments
Easy Protocol Setup
  • Offers a wide array of plate templates from major manufacturers, simplifying the setup process.
Excel Integration
  • Efficient Data Management
Enhanced Calibration Capabilities
  • Includes precalibrated liquid classes and guided wizard
Specifications:
User Interface
On-board interface or FILLit PC software
Volume
0.5-2500 µl 
Volume increments 
Small tube dispensing cassette: 0.5–50 μL, in 0.5 μL increments
Standard tube dispensing cassette: 5- 2,000 µL in 5 µL increments
Dispensing technology
Peristaltic pump
Dispensing volume adjustability
Per column
Dead Volume
< 1 mL volume range: 0.5 - 50 µL
<7 mL volume range: >50 µL
(it is possible to backflush the reagent into original reservoir to avoid loss of expensive reagent)
Plate Format
9- to 1,536-well plates: including 96- and 384-deep well and tubes in rack format (plate height: 5 - 50 mm)
Automation/robot compatible
Yes
Dimensions
35.6 x 21.8 x 32.8 cm
Weight
9.1Kg
Ordering Information
Order below or select product to learn more.
Catalog # Name Size Price Qty
5840330 Each
21,240.90
Online Exclusive
24,191.00 Save 2,950.10 (12%)
5840340 Each
26,010.90
Online Exclusive
29,624.00 Save 3,613.10 (12%)
5840400 Each
37,412.72
Online Exclusive
43,023.00 Save 5,610.28 (13%)
For Research Use Only. Not for use in diagnostic procedures.