Custom TaqMan Plates are cost-effective and convenient 96-well plates containing dried-down TaqMan Gene Expression Assays. Customize any of the plate formats to fit your experimental design. The flexibility enables an efficient means to screen biological samples for the presence and amount of specified gene targets expressed in any species covered in our collection of over 2.3 million predesigned TaqMan Gene Expression Assays. You can select from 9 plate formats in either a Standard 0.2 mL MicroAmp Optical 96-well Reaction Plate (20 μL final reaction volume) or a Fast 0.1 mL MicroAmp Optical 96-well Reaction Plate (10 μL final reaction volume). It’s easy to design a TaqMan Array Plate with the assays you want, in the arrangement you need.
  • Customizable—use our new online ordering tool to select assays, replicates, controls, and more
  • Affordable—order as few as six plates
  • Efficient—setup in less than 10 minutes
  • Convenient—just add master mix and your cDNA sample
  • Broad instrument compatibility—compatible on all Applied Biosystems qPCR instruments; Fast 0.1 mL plates also compatible with BioRad™ CFX Connect™ and CFX96 Touch™ instruments
Adding controls
18S can be chosen as a control for human, mouse and rat assays. GAPDH can only be chosen as a control for human assays. All TaqMan Array Plate Plus* formats include three human endogenous control genes: GAPDH, HPRT, and GUSB, which are expressed at relatively abundant levels across numerous tissues (based on in-house research). TaqMan Array Plate Plus* formats should only be used for experiments with human assays, not any other species.
TaqMan® Array Standard Plates
20 µL reaction
Order Now
TaqMan® Array Fast Plates
10 µL reaction
Order Now
Format*
Catalog #
(Fast)
Catalog  # (Standard) # of Assays/ Controls # of Samples Min. Order
TaqMan® Array Plate 8 4413263 4413266 7 + 1 12  6
TaqMan® Array Plate 16 4413261 4413264 15 + 1 6  6
TaqMan® Array Plate 16 Plus 4413262 4413265 12 + 4 6  6
TaqMan® Array Plate 32 4413259 4391528 31 + 1 3  6
TaqMan® Array Plate 32 Plus 4413260 4391529 28 + 4 3  6
TaqMan® Array Plate 48 4413257 4391526 47 + 1 2  6
TaqMan® Array Plate 48 Plus 4413258 4391527 44 + 4 2  6
TaqMan® Array Plate 96 4413255 4391524 95 + 1 1  6
TaqMan® Array Plate 96 Plus 4413256 4391525 92 + 4 1  6
 * TaqMan® Array Plus plates only use Human Endogenous Controls, controls for no other species are used.