Natriuretic-Peptide-Receptor-C_Antibody-470x200
We offer a wide range of peptides and recombinant proteins for use in your experiments. Our custom peptides and peptide libraries are available with flexible modification, conjugation, and isotopic labeling options.
For applications including proteomics, biomarker discovery, antibody production, and more, our peptide synthesis service can help you design and synthesize reliable and validated peptides. Our recombinant proteins can be used as standards and controls in cell assays and immunoassays. In addition, we offer function, high-quality, purified growth factors for use in cell culture.
Featured peptide & protein categories
Our Standard Peptide Synthesis Service offers custom peptides with over 400 modifications options available to help meet a variety of research needs. Peptides between 2 and 110 amino acids in length, a range of scales (1 mg–1 g) and purity (crude to >98%) are available.
The HeavyPeptide AQUA Custom Synthesis Service provides isotopically labeled, AQUA (Absolute QUAntitation)-grade peptides for the relative and absolute quantitation of proteins.
PEPotec SRM Custom Peptide Libraries are optimized for selected or multiple reaction monitoring (SRM/MRM) methods of absolute quantitation by mass spectrometry.
A broad range of proteins, including functional growth factors, recombinant and native proteins; select from our Invitrogen recombinant proteins or from the portfolio of Sino Biological.
Designed specifically for the identification of immunogenic protein sites, these custom crude peptide libraries feature a low-toxicity formulation.
Explore our collection of high-quality proteins for use as growth factors and cell-signaling molecules, such as cytokines and chemokines.
Resources