Choose the appropriate dialysis membrane cut-off and device size
The use of dialysis to separate complex mixtures of biomacromolecules was established in the 1950s. Many of the dialysis theories established at that time are the cornerstones for contemporary dialysis products. However, modern dialysis tools differ from those of the past in two key respects—preparation time and sample recovery. While early laboratory dialysis methods involved the dedication of a significant amount of time to membrane preparation, the Thermo Scientific Pierce Dialysis Products described on this page are essentially ready to use and resist sample leakage.
SnakeSkinBeaker190x233-sc SnakeSkin Dialysis Tubing
SnakeSkin™ Dialysis Tubing
SnakeSkin Dialysis Tubing was developed to simplify large-volume sample dialysis and does not require presoaking or boiling the membrane prior to use. SnakeSkin Dialysis Tubing is regenerated cellulose dialysis tubing that is pleated (compressed) into a hollow stick and can be used for a wide range of sample volumes. The hydrated tubing will hold ~3.7 mL of sample per centimeter of length.
The pleated format of SnakeSkin Dialysis Tubing makes it easy to open and ready to use; simply pull out the required length of tubing, cut it off and apply a closure. The recommended closures, SnakeSkin Dialysis Tubing Clips, are sold separately. Alternatively, SnakeSkin Dialysis Tubing can also be closed with knots after prewetting the tubing in water or buffer.
SnakeSkin Dialysis Tubing is available in three molecular weight cutoff (MWCO) specifications: 3.5K, 7K and 10K.
High-Performance Dialysis Brochure
Dialysis is a separation technique that gained popularity in life science laboratories during the 1950s. Research papers of that era described dialysis as a new, cutting-edge tool that scientists could use to unravel complex mixtures of biomacromolecules. Many of the dialysis theories established at that time are the cornerstones for contemporary products featured in this brochure. There are, however, two major differences between the dialysis tools of yesterday and today—preparation time and the amount of sample loss due to leaks. Early laboratory dialysis methods involved dedicating a significant amount of time to membrane preparation; Thermo Scientific Pierce Dialysis Products are essentially ready to use and resist sample leakage.
Sal-Flask-190x250 Slide-A-Lyzer Dialysis Flasks
Slide-A-Lyzer Dialysis Flasks
Slide-A-Lyzer Dialysis Flasks are for sample volumes from 150 to 250 mL. These devices accomplish typical buffer exchange in 8 to 24 hours and are available in four molecular-weight cutoff (MWCO) varieties: 2K, 3.5K, 10K, and 20K.
The flask-style dialysis devices are very easy to use. Add and remove sample using a serological pipette or by directly pouring the sample through a wide screw-cap opening at the top of the flask. No knots or clips are needed to seal the units.
Slide-A-Lyzer Dialysis Flasks are manufactured using clean room conditions and have low-protein binding regenerated cellulose membranes to ensure maximum sample recovery and sample purity.
SalBeaker190x251 Dialysis Cassettes
SalG2Composite190x251 G2 Dialysis Cassettes (self-floating with pipette port)
Slide-A-Lyzer Dialysis Cassettes
Slide-A-Lyzer Dialysis Cassettes (original and G2 models) facilitate rapid and trouble-free dialysis for sample volumes from 100 µL to 70 mL. The cassette design maximizes surface-area per sample-volume ratio and allows excellent sample recoveries. Unlike standard flat tubing, these innovative cassettes do not require knots or clips that can lead to leaking and sample loss. Instead, Slide-A-Lyzer Dialysis Cassettes are constructed of dialysis membranes separated by an inert gasket and sandwiched between two halves of a translucent plastic case that is sonically welded together.
  • Original-style Dialysis Cassettes:
    Designed with an inner chamber that is hermetically sealed, allowing the sample to be accessed by inserting a syringe needle through the inlet port.
  • 2nd-Generation (G2) Dialysis Cassettes:
    Designed with two additional features: (1) they have needle-accessible ports plus a unique pipette-accessible port, and (2) they are self-floating.
  Original
Slide-A-Lyzer
Cassettes
G2
Slide-A-Lyzer
Cassettes
MWCOs offered Five (2 to 20K) Five (2 to 20K)
Sizes offered Four (0.1 to 30 mL) Five (0.1 to 70 mL)
Syringe injection ports Yes Yes
Pipette sample port No Yes
Self-floating 30 mL size only Yes
Both models of Slide-A-Lyzer Dialysis Cassettes are available in five molecular weight cut-off (MWCO) specifications (2K, 3.5K, 7K, 10K, and 20K) for four or five different volume ranges. Selected sizes of 10K MWCO Slide-A-Lyzer Cassettes of each model are also available in packages that have been gamma-irradiated to make them effectively sterile.
Guide to available Slide-A-Lyzer Dialysis Cassettes (SAL and SAL G2 models).
Selecting a link based on desired molecular weight cut-off (MWCO) and cassette size (sample volume) opens the product page.
Sample Volume 2K MWCO
(blue)
3.5K MWCO
(pink)
7K MWCO
(green)
10K MWCO
(orange)
20K MWCO
(purple)
0.1–0.5 mL SAL SAL SAL SAL SAL
SAL G2 SAL G2 SAL G2 SAL G2 SAL G2
0.5–3 mL SAL SAL SAL SAL SAL
SAL G2 SAL G2 SAL G2 SAL G2 SAL G2
3–12 mL SAL SAL SAL SAL SAL
5–15 mL SAL G2 SAL G2   SAL G2 SAL G2
12–30 mL SAL SAL   SAL SAL
SAL G2 SAL G2   SAL G2 SAL G2
30–70 mL SAL G2 SAL G2   SAL G2 SAL G2
MiniSalComposite190x204 Slide-A-Lyzer MINI Dialysis Devices and Floats
Slide-A-Lyzer MINI Dialysis Devices
Slide-A-Lyzer MINI Dialysis Devices are disposable polypropylene cups with integrated, low-binding membranes for convenient, high-performance dialysis of 10 µL to 2 mL samples. The MINI devices are designed to allow easy sample addition and removal using a standard laboratory pipette. They are effective and convenient for accomplishing typical sample desalting or buffer exchange in 4 to 8 hours. Unlike other small sample separation devices, there is no equipment to assemble, no need for syringe adapters and no laborious steps necessary to manipulate the small volume sample. The regenerated cellulose membrane is compatible with a number of common chemicals and buffers.
Slide-A-Lyzer MINI Dialysis Devices are available in three sizes (0.1, 0.5 and 2 mL capacities) with five molecular weight cut-off (MWCO) specifications: 2K, 3.5K, 7K, 10K, and 20K.
Slide-A-Lyzer MINI Dialysis Devices—specific product page links
Microdialysis-plates190x190 96-well Microdialysis Plates
Pierce 96-well Microdialysis Plates
Pierce 96-well Microdialysis Plates are automation-compatible, microplate dialysis devices (3.5K or 10K MWCO) for rapidly processing 1 to 96 samples with volumes 10 to 100 µL.
Each sample processed with the Pierce 96-well Microdialysis Dialysis Plate is dialyzed inside a single, removable dialysis device created by two regenerated cellulose membranes separated by ~2 mm. The short diffusion distance and large surface area facilitates rapid dialysis with the removal of salts and small molecules often completed in 2 to 4 hours. Sample recovery is performed using standard laboratory micropipettes. Individual microdialysis devices can also be used with 2 mL microcentrifuge tubes for processing single samples. The available plate seals are made of high-quality polypropylene and help protect samples from contamination and evaporation, especially during long-term storage.
RedRakefingerinsert190x189 RED Device Rapid Equilibrium Dialysis System
RED Device for rapid equilibrium dialysis
The RED Device for rapid equilibrium dialysis was developed in association with the pharmaceutical industry to accelerate lead optimization and reduce attrition rate. The RED System consists of disposable tube inserts for use with either a reusable PTFE or disposable high-density polypropylene base plates. The unique design of the base plates provides compatibility with automated liquid handling systems while the large dialysis surface area of the tube inserts accelerates equilibrium establishment.
C-REDComposite190x172 Competition Rapid Equilibrium Dialysis System
Competition Rapid Equilibrium Dialysis System
Like the RED Device, the Competition Rapid Equilibrium Dialysis System was developed with accelerated lead optimization as a priority. The Competition RED Device (C-RED) consists of two-chambered disposable tube inserts and a reusable PTFE base plate. Competition equilibrium dialysis is performed within enlarged wells on the base plate that can accommodate 2, 3, 4, or 8 dual chamber inserts providing a defined space for up to 15 samples and 1 open chamber to access the dialysate. The Competition Rapid Equilibrium Dialysis System is also compatible with automated liquid handling systems and has the same high performance as the RED Device.