Search
The Ion AmpliSeq Rapid Microbial Identification (RMID) Panel is a pre-designed custom targeted next-generation sequencing (NGS) research panel. It uses Ion AmpliSeq technology for targeted NGS to research bacterial 16S rRNA, fungal rRNA, and rpoB regions, allowing for pan-bacterial and pan-fungal metagenomic detection, along with species-specific identification of 88 microbial species in a single assay.
The panel supports DNA extracted and purified from various research sample types, including FFPE tissue, bronchoalveolar lavage (BAL), cerebrospinal fluid (CSF), blood, joint fluid, and stool. This analysis can be completed in as little as 24 hours with automated NGS.
| Targets for species-specific identification | ||||
| Gram-negative bacteria | Desulfovibrio alaskensis | Phascolarctobacterium faecium | Collinsella stercoris | Peptostreptococcus anaerobius |
| Acinetobacter baumannii | Enterobacter cloacae | Porphyromonas gingivalis | Dorea formicigenerans | Peptostreptococcus stomatis |
| Bacteroides fragilis | Escherichia coli | Prevotella copri | Enterococcus faecalis | Propionibacterium acnes |
| Bacteroides nordii | Fusobacterium nucleatum | Prevotella histicola | Enterococcus faecium | Roseburia intestinalis |
| Bacteroides thetaiotaomicron | Gardnerella vaginalis | Proteus mirabilis | Enterococcus gallinarum | Ruminococcus bromii |
| Bacteroides vulgatus | Gemmiger formicilis | Pseudomonas aeruginosa | Enterococcus hirae | Ruminococcus gnavus |
| Barnesiella intestinihominis | Haemophilus influenzae | Serratia marcescens | Eubacterium limosum | Slackia exigua |
| Borrelia burgdorferi | Helicobacter bilis | Veillonella parvula | Eubacterium rectale | Staphylococcus aureus |
| Campylobacter concisus | Helicobacter bizzozeronii | Gram-positive bacteria | Faecalibacterium prausnitzii | Staphylococcus epidermidis |
| Campylobacter curvus | Helicobacter hepaticus | Anaerococcus vaginalis | Holdemania filiformis | Staphylococcus haemolyticus |
| Campylobacter gracilis | Helicobacter pylori | Atopobium parvulum | Lactobacillus acidophilus | Streptococcus gallolyticus |
| Campylobacter hominis | Klebsiella oxytoca | Bifidobacterium adolescentis | Lactobacillus delbrueckii | Streptococcus infantarius |
| Campylobacter jejuni | Klebsiella pneumoniae | Bifidobacterium animalis | Lactobacillus johnsonii | Streptococcus pneumoniae |
| Campylobacter rectus | Mycoplasma fermentans | Bifidobacterium bifidum | Lactobacillus murinus | Streptococcus pyogenes |
| Chlamydia pneumoniae | Mycoplasma penetrans | Bifidobacterium longum | Lactobacillus reuteri | Streptococcus salivarius |
| Chlamydia trachomatis | Neisseria meningitidis | Blautia obeum | Lactobacillus rhamnosus | Streptococcus sanguinis |
| Citrobacter freundii | Parabacteroides distasonis | Clostridium difficile | Lactococcus lactis | Yeast |
| Citrobacter rodentium | Parabacteroides merdae | Collinsella aerofaciens | Parvimonas micra | Candida albicans |
Dr. Jennifer Dien Bard discusses her experience using an automated targeted NGS method to rapidly detect and identify bacteria and fungi directly from specimens.
Figure from Osborn LJ et al. J Clin Microbiol. 2024 May 8;62(5):e0174923, used with permission.
The Ion AmpliSeq RMID Panel is a custom targeted NGS panel. You can use the pre-designed panel as-is or customize to adapt to your unique NGS testing needs. Ion AmpliSeq panels are based on ultrahigh-multiplex PCR, enabling clinical microbiology researchers to rapidly and easily detect and identify bacterial and fungal species in samples.
When navigating through the complexities of infectious disease and microbiology research, you need a discovery tool that is both flexible and rapid for analyzing microbes of interest. Ion AmpliSeq technology can help. It is globally recognized as a widely adopted amplicon-based enrichment method for targeted NGS. Clinical and translational researchers worldwide choose Ion AmpliSeq technology for a fast, simple, and scalable NGS solution.
Ion AmpliSeq Designer is an online primer design tool that lets you create custom panels for Ion Torrent NGS. Available in ready-to-use, made-to-order, and on-demand formats, Ion AmpliSeq panels consist of a pool of oligonucleotide primer pairs, with each pair designed to amplify a specified genomic region. Each panel can be designed to interrogate entire microbial genomes or all bases across a gene, or it can be focused on specific regions to study microbial variants or mutation hotspots.
The Ion AmpliSeq RMID Panel is compatible with both the Ion GeneStudio S5 systems and the Genexus System to deliver ease of use, automation capabilities, and rapid NGS test results in as little as 24 hours.
Through a combination of workflow automation, sequencing performance, and automated data analysis and reporting, the Ion Torrent Genexus System brings the power of in-house NGS to microbiology labs worldwide without the need for extensive sequencing and bioinformatics expertise.
Helping clinical microbiology researchers achieve deeper insights through fast, efficient, and cost-effective generation of large sequencing datasets, the Ion GeneStudio S5 systems enable scalable, high-throughput NGS for a broad range of applications.
We will be happy to answer your questions and provide a demo of our rapid microbial identification (RMID) NGS solutions.
For Research Use Only. Not for use in diagnostic procedures.
CN: 62894