Performance Specifications Q2 2012 2H 2012
Input DNA 10 ng -
Cumulative target bases 1 kb to 250 kb 1 kb to 1 Mb
Multiplexity (primer pairs/tube) 12 to 1,536 12 to 3,072
in silico base coverage >85% -
On-target bases, specificity >80% -
Typical coverage uniformity (>0.2x mean) 85% 95%-100%
Support
Please contact Ion Torrent™ at: 
1-87-SEQUENCE (1-877-378-3623) 
Email the Support Team >
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.