Search
In the world of quality control, precision and accuracy are paramount. Ensuring the best quality of the final product is of utmost importance and, to reach the highest standards, industries from various fields make use of different analytical techniques. These techniques are used to assess and monitor the quality of products or materials and the choice of a specific technique employed varies depending on the industry, the nature of the products being tested, and the information needed to ensure the product quality.
Most analytical techniques employed in quality control and assurance, such as inductively coupled plasma-optical emission spectroscopy (ICP-OES), are meant to provide general information about the composition of the material such as chromatography, or about possible trace elements. These techniques can have key roles in assessing physical properties. For example, thermal stability is assessed with differential scanning calorimetry (DSC) and thermogravimetric analysis (TGA).
However, energy-dispersive X-ray spectroscopy (EDS) has proven to be invaluable for its speed, accuracy, and versatility. Compared to the above-mentioned techniques, EDS provides insights into the composition and characteristics of materials of specific areas of interest allowing to find defects, contaminants, and to characterize the surface morphology. EDS can result in reduced materials waste, reduced costs, and reduced number of faulty and defective items. Additionally, because production is more controlled and monitored at every step of the EDS process, the overall quality of the final product is better.
EDS characterization of unknown materials on a face mask
EDS is primarily used to verify the presence and distribution of specific elements in a sample. EDS is based on the analysis of the characteristic X-rays emitted by the sample after interacting with a primary electron beam from the scanning electron microscope (SEM) source. The analysis and processing of the characteristic x-rays provides the elemental composition across an area of interest and, while it is crucial to characterize the expected elements in a material, is even more important to identify the presence of possible impurities or contaminants. This information ensures that materials meet any required specifications and standards.
In fact, contamination detection is one of the main uses for EDS in quality control as the presence of impurities, the identification of their nature and origin are a major concern in quality control. EDS not only excels in the detection of contaminants, even at trace levels, but it provides information also about the localization of the unknown material in the product. This allows users to determine which stage the impurity entered the production process or if it comes from the initial raw material. Being able to trace the origin of a contaminant enables users to take corrective measures, prevent future occurrences and, eventually, improve the material selection.
In the event of product failure or customer complaints, EDS also plays a crucial role in failure analysis. With the combined use of SEM-EDS, manufacturers can investigate and identify the root causes of failures or defects. EDS helps determine whether the failure was caused by material composition issues, contamination, or other factors. This information helps resolve customer complaints, improves product design, and helps implement corrective actions to prevent similar future failures.
Quality control and assurance are not limited to assessing finished products. EDS also supports research and development processes by providing valuable insights in the design and optimization phase. Researchers can use EDS to explore new materials, study their behavior, and assess their performance. By gaining a deeper understanding of the elemental composition and structure of materials, companies can develop innovative products that meet the evolving demands of their customers.
The use of EDS in quality control and assurance processes offers numerous advantages to manufacturers across varying industries. From accurate elemental analysis and non-destructive testing to enhanced defect detection and material verification, EDS enables companies to ensure the production of high-quality products. By incorporating this advanced technique into their quality management systems, industries in various fields can enhance their product quality, optimize manufacturing processes, and ultimately improve customer satisfaction.
For Research Use Only. Not for use in diagnostic procedures.