Iridium • Transition Metal
Primary XPS region: Ir4f
Overlapping regions: N/A
Binding energies of common chemical states:
Chemical stateBinding energy Ir4f7/2
Ir metal60.9 eV
Experimental information
N/A
Interpretation of XPS spectra
  • Ir4f region has well separated spin-orbit components (Δ=3.2eV).
    • Peaks in the Ir4f region have an asymmetric peak shape for iridium metal.
    • Iridium compounds, such as the oxide, have symmetric Ir4f peaks.
About this element
Element Crystal fcc
Symbol: Ir
Date of discovery: 1803
Name origin: Greek iris
Appearance: white
Discoverer: Smithson Tennant, et al.
Obtained from: deposits with platinum
Melting point: 2739 K
Boiling point: 4701 K
Density[kg/m3]: 22650
Molar volume: 8.52 × 10-6 m3/mol
Protons/Electrons: 77
Neutrons: 115
Shell structure: 2,8,18,32,15,2
Electron configuration: [Xe]4f145d76s2
Oxidation state: 2,3,4,6
Crystal structure: cubic face centered
Iridium is used as a hardening agent for platinum and for making tools that are exposed to high temperatures, like crucibles. Iridium salts vary widely in color, which is why iridium was named after the Latin word for rainbow: iris. The KT extinction event was a period over 65 billion years ago that marked the extinction of many forms of life such as, most popularly, the dinosaurs. This time period that marks the temporal border between the Cretaceous and Tertiary eras in geological time was identified by a thin layer of clay that contained an abnormally high level of iridium. Many scientists believe that iridium originated from an asteroid or comet that was the cause of the extinction of the dinosaurs. However, because the Earth’s core is rich in iridium, other scientists argue that iridium came from volcanic origin.
Join-the-conversation-banner-in-blogs-social-graphic-1143250-1000x172
element_iridium
Products
Style Sheet for Instrument Cards Original
K-Alpha XPS
  • High resolution XPS
  • Fast, efficient, automated workflow
  • Ion source for depth profiling
ESCALAB QXi XPS
  • High spectral resolution
  • Multi-technique surface analysis
  • Extensive sample preparation and expansion options
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Electron microscopy services for
the materials science
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.