Technical Resources - Media Formulations
51300 - Insulin, Transferrin, Selenium, Sodium Pyruvate Solution (ITS-A)
ComponentsMolecular WeightConcentration (mg/L)mM
Proteins
Insulin5808.01000.00.1721763
Transferrin80000.0550.00.006875
Trace Elements
Sodium Selenite173.00.670.0038728325
Other Components
Sodium Pyruvate110.011000.0100.0
参照