Reduce your design cycle, inspection times, and meet higher-level quality standards at a lower cost
Imaging techniques such as computed tomography are increasingly used in industry as a method for nondestructive testing, adding to standard CMM the ability to make digital measurements and detect and quantify defects inside a part.
We developed Thermo Scientific Avizo Software in collaboration with major industrial partners in the aerospace, automotive, and consumer goods industries. We have integrated expertise acquired over more than 10 years so that you can use Avizo Software to visualize, analyze, and inspect parts and materials acquired by CT systems and microscopy. 
Avizo Software provides a comprehensive set of tools that address the entire research-to-production cycle: from materials research in off-line labs to automated quality control in production environments. With Avizo Software, you can reduce your design cycle and inspection times, while meeting higher-level quality standards at a lower cost.
Imaging data
Workflow using Avizo Software
Processing
Workflow using Avizo Software
Visualization
Workflow using Avizo Software
Analysis
Workflow using Avizo Software
When designing a new mechanical part, prototyping its manufacturing process, monitoring quality on the production line, or inspecting it after it fails, X-ray CT combined with software applications allows you to optimize your design, increase production quality, and quickly fix a possible design flaw.
Avizo Software allows for detection and quantification of porosity, distribution, clustering, closest pore, distance to surface of the part), corrosion, cracks in metal alloys or polymer parts, delamination, gluing issues, change of density, or voids in the matrix in a composite material part, for instance.
Avizo Software can measure wall thickness and import CAD models to perform surface deviation analysis.
Avizo defect analysis
Defect analysis on compressor housing. Data courtesy of CyXplus.
While digital industrial inspection aims at improving production quality, the inspection process should also be able to provide cost reductions and support increased productivity. The ultimate goal is to fully automate complex inspection scenarios.
Avizo Software for industrial inspection provides an advanced recipe mechanism that allows defect detection and part characterization to be automated. For a particular inspection workflow, the recipe is created just once and can then be replayed at any time. The recipe includes advanced analysis processes that give meaningful insight about defects that are detected during the inspection. The automatically generated quantifiable measurements allow a proper assessment of the part’s quality. .
While digital industrial inspection aims at improving production quality, the ability to document the results generated during the inspection workflow and maintain a proper traceability of these results is of utmost importance.
Avizo Software for industrial inspection provides reporting and traceability mechanisms. The generation of these reports can even be automated through the recipe functionality. Reports can include 3D visualization snapshots of the generated results, as well as spreadsheets containing the results of the different analyses. Each result generated through the inspection workflow contains a history log describing the different steps leading to this particular result.
Complex inspection scenarios, combining multiple recipes on different sub-regions of the part or material, can be created using Avizo Software (in-line version). Scenarios can combine multiple steps of visual inspection and automatic or manual measurements. Inspector services run scenarios created with Avizo Software designer and will accept or reject a part according to automated or interactive acceptance and/or tolerance values associated with automatic and/or manual measurements.
Avizo Software directly connects Inspector services with the acquisition system, streamlining the inspection process. Avizo Software (in-line version) pre-processes data pushed to a DICOM/DICONDE server; performs pre-processing, such as multi-part splitting, part registration to reference model, and acquisition artifact reduction; then dispatches the pre-processed part to a scalable collection of Inspector services. At any time, the results of inspection scenarios can be accessed with Reviewer services, where each step of the scenario can be reviewed and a final decision on part acceptance made. Avizo Software also includes user administration, allowing assignment and management of different permission levels.
Avizo_Industrial-inline-integration
Techniques
Services
Amira-Avizo-Introductory-training_1160x600
Introductory training
Shorten your learning curve and maximize your investment with this introductory training specifically designed for new users of Amira, Avizo and PerGeos Software.
The course consists of a lecture with hands-on sessions. The training material highlights the basic features and functionalities of Amira, Avizo and PerGeos Software.
Amira-Avizo-advanced-training_1160x600
Advanced training
Maximize your investment and reduce your time-to-results with this advanced training specifically designed for existing users of Amira, Avizo and PerGeos Software.
The course consists of a lecture with hands-on sessions. The training material highlights advanced features and functionalities of Amira, Avizo and PerGeos Software.
Amira-Avizo-Custom-Dev_1160x600
Custom development
With over 25 years of experience in 3D and image processing and hundreds of custom projects delivered to organizations small and large, Thermo Fisher Scientific can provide you with a solution tailored to fit your specific needs.
We can customize and expand our software solutions at various levels.
Resources
Avizo for Industrial Inspection - Overview
Discover Avizo Software solutions for quality control
Avizo for Industrial Inspection - Easy creation & customization of inspection workflows
Avizo for Industrial Inspection - Reporting & traceability
Multiscale Image-based control and characterization white paper
Multiscale Image-Based Control and Characterization
Enhance batteries performance and increase safety
Read more
Avizo Software brochure
Avizo Software brochure
Avizo Software allows for a better understanding of structure, properties and performance.
Read more
Avizo for Industrial Inspection - Overview
Discover Avizo Software solutions for quality control
Avizo for Industrial Inspection - Easy creation & customization of inspection workflows
Avizo for Industrial Inspection - Reporting & traceability
Multiscale Image-based control and characterization white paper
Multiscale Image-Based Control and Characterization
Enhance batteries performance and increase safety
Read more
Avizo Software brochure
Avizo Software brochure
Avizo Software allows for a better understanding of structure, properties and performance.
Read more
Features
Import and process your imaging data
    • Handle any modality, at any scale, of any size:
    - Bio-Formats
    - Bitmap formats
    - Microscopy: electron and optical
    - Medical and neuroimage formats
    - Molecular formats
    - Other acquisition devices (MRI, radiography, etc.)
    • Finite element modeling, geometric modeling, CAD
    • Support for multi-data/multi-view, multi-channel, time series, very large data
    • Scaling, calibration, conversion, re-sampling
    • Image enhancement, comprehensive filtering and convolution, Fourier frequency transforms
    • Artifact reduction algorithms
    • Advanced multi-mode 2D/3D automatic registration
    • Image stack alignment, arithmetic, correlation, fusion
    Easily segment your imaging data
    • Thresholding and auto-segmentation, object separation, automatic labeling
    • Region growing, snakes, interpolation, wrapping, smoothing
    • Morphological processing, including watershed and basins
    • Machine Learning-based segmentation
    • Automatic tracing of individual fibers and filaments
    • Skeletonization and filament network extraction
    • Interactive tools for generation or editing of segmentation and spatial graphs
    • 3D surface reconstruction
    • Grid generation for FEA/CFD
    Export your analysis and visualization work to seamlessly publish and present it
    • Animation and video generation
    • Advanced key frame and object animation
    • Mix images, geometric models, measurements and simulations
    • Annotations, measures legends, histograms and curve plots
    • Export spreadsheets, 3D models, high-quality images
    • Active and passive 3D stereo vision
    • Single and tiled screen display
    • Immersive environment
    Visualize and explore your imaging data
    • Interactive high-quality volume and multichannel visualization
    • Orthogonal, oblique, cylindrical and curved slicing
    • Contouring and iso-surface extraction
    • Maximum Intensity or other types of projections
    • Vector and tensor visualization
    • Objects and tracks
    • Molecular visualization
    Analyze your imaging data and obtain quantitative information
    • Intuitive recipe creation, customization, automated replay
    • Built-in measurements, including counts, volumes, areas, perimeters, aspect ratios and orientations
    • User-defined measures
    • Results viewer with spreadsheet tool and charting
    • Automatic individual feature measurements, 3D localization and spreadsheet selection
    • Automated statistics, distribution graphs
    • Feature filtering using any measurement criterion
    • Data registration, deformation, comparison and measurements
    Easily and quickly adapt Amira Software to your specific needs
    • Custom C++ modules development
    • MATLAB™ bridge
    • Python scripting API
    Contact us