Tungsten • Transition Metal
Primary XPS region: W4f
Overlapping regions: N/A
Binding energies of common chemical states:
Chemical stateBinding energy W4f7/2
W metal31.6 eV
WS232.4 eV
WO233.1 eV
WO336.1 eV
Sulfide and oxides charge referenced to adventitious C1s peak at 284.8eV.
Experimental information
N/A
Interpretation of XPS spectra
  • W4f region has well-separated spin-orbit components (Δmetel=2.17eV).
    • Loss feature for W metal appears at 37eV and 42eV for WO3. W metal loss feature overlaps WO3 4f peaks.
    • Peaks in the W4f region have an asymmetric peak shape for tungsten metal.
    • Tungsten compounds, such as the oxide, have symmetric W4f peaks.
  • Use Smart background when sputter profiling films such as WO3 on W.
    • Linear or Shirley background which is suitable for WO3, for example, will not be suitable for W metal.
w4f-spectrum-native-oxide-w
General comments
  • Tungsten oxide is reduced by an argon ion beam to form a continuum of oxidation states.
    • Use lowest beam energy possible for depth profiling or cluster source to clean (if available).
About this element
Element Crystal bcc
Symbol: W
Date of discovery: 1783
Name origin: Swedish tung sten
Appearance: silver
Discoverer: F. and J. J. de Elhuyar
Obtained from: scheelite, wolframite
Melting point: 3695 K
Boiling point: 5828 K
Density[kg/m3]: 19250
Molar volume: 9.47 × 10-6 m3/mol
Protons/Electrons: 74
Neutrons: 110
Shell structure: 2,8,18,32,12,2
Electron configuration: [Xe]4f145d46s2
Oxidation state: 6,5,4,3,2
Crystal structure: cubic body centered
Tungsten’s symbol, W, originates from its former name "wolfram". Tungsten is found in ores including wolframite, which was named after Peter Wolfe who first hypothesized the existence of tungsten in wolframite. It is extremely resistant to corrosion and forms a protective oxide when exposed to air. The largest use for tungsten is as tungsten carbide used in cemented carbides (wear-resistant materials used in metal working, mining, petroleum, and construction industries). Because it can be made into very thin wires and due to its high melting point, tungsten is applicable in light bulbs, vacuum tube filaments, and electron guns. Tungsten salts are also used in the tanning industry.
element_tungsten
Products
Nexsa XPS
  • Módulo de basculación para mediciones de ARXPS
  • Fuente de iones de modo dual para capacidades de realización de perfiles de profundidad
  • Análisis de aislantes.
Hoja de estilo para tarjetas originales instrumentos
K-Alpha XPS
  • Espectroscopía de área seleccionable.
  • Monocromador microfocalizado.
  • Espectroscopía del estado químico de alta resolución.
ESCALAB Xi+ XPS
  • Espectroscopía de alta sensibilidad.
  • XPS con fuente de rayos X no monocromática.
  • Analizador hemisférico de 180°.
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Hoja de estilo para las especificaciones de la tabla de productos
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Hoja de estilo para las especificaciones de la tabla de productos
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Servicios de microscopía electrónica para
la ciencia de materiales
Para garantizar un rendimiento óptimo del sistema, le proporcionamos acceso a una red de expertos de primer nivel en servicios de campo, asistencia técnica y piezas de repuesto certificadas.