HeLa Cells labeled with 2 µg/mL HCS CellMask™ Blue Stain
Fixed and permeabilized HeLa Cells labeled with 2 µg/mL HCS CellMask™ Blue Stain (Cat. No. H32720) for 30 minutes at room temperature. Segmentation performed on the Cellomics ArrayScan® VTI (left image).
Fixed and permeabilized HeLa Cells labeled with 2 µg/mL HCS CellMask™ Blue Stain (Cat. No. H32720) for 30 minutes at room temperature.  Segmentation performed on the Cellomics ArrayScan® VTI (left image).
Related Products
Related Images