The symptoms are caused by the deposition of amyloid fibrils in tissues. The fibrils are formed from monoclonal light chains. Worldwide incidence is about 9 cases per million per year 1 with a prevalence of 58 cases per million 2.
Unlike multiple myeloma (MM), the morbidity and mortality associated with AL amyloidosis are due to the accumulation of FLC-derived amyloid fibrils in the organs, and not directly due to the proliferation of monoclonal plasma cells. This deposition may occur in a number of different organs including the heart, kidney, liver and nervous system 3.
The kidney is the organ most frequently involved, with 70-80% of AL amyloidosis patients having renal involvement 4. The heart is the next most common site of involvement, with 50-60% of patients having evidence of cardiac involvement by echocardiography 4, followed by liver involvement (25%) and involvement of the peripheral nervous system in 20% of AL amyloidosis patients 4. It is particularly important to identify cardiac involvement so treatment of the underlying monoclonal gammopathy can begin alongside supportive care for the cardiac dysfunction, as 75% of deaths in AL amyloidosis are due to heart failure or arrhythmia caused by the amyloid fibril deposits 5.
For this reason, a variety of clinical and laboratory tests are used to diagnose or rule out AL amyloidosis 7.
Once identified, a number of treatment options are available for AL amyloidosis patients - these can include drugs that target the underlying tumor (the source of the amyloidogenic FLC) and stem cell transplant.
Response to treatment in AL amyloidosis is assessed by organ response and hematological response. Hematological response is evaluated by measurement of free light chains 9.
In AL amyloidosis, the dFLC (difference between the involved FLC and uninvolved FLC) is used in response assessment.
It is very important to assess hematological response, because it begins to occur earlier than organ response, and can tell clinicians whether a treatment is working or whether it needs to be changed.
Around 10-15% of patients with multiple myeloma may develop concurrent AL amyloidosis 10. It’s important to be aware of this when managing patients with myeloma, and be alert for signs of amyloid deposit-related organ damage so supportive care given if needed.
Haematological response and progression criteria 9 | ||||
---|---|---|---|---|
Complete response | Normal FLC ratio, and Negative IFE of serum and urine, and <5% plasma cells in bone marrow | |||
Very good partial response | Reduction in dFLC to <40mg/L | |||
Partial response | >50% decrease in dFLC | |||
No response | Less than a PR | |||
Progression | From CR | Detectable M-protein or abnormal sFLC ratio (FLC must double) | ||
From PR | increase in urine M-protein to >200mg/day |