Over the past two decades there has been a decisive shift towards large molecule, biological drugs or biopharmaceuticals, and away from traditional chemically synthesised, small molecule drugs. The reasons for this, I believe, are outlined in a previous article entitled Pharma Blockbuster League: Biopharmaceuticals 7 – Small Molecules 3. This is a trend which nearly all commentators agree will continue, but which biomolecules is the pharmaceutical industry currently using for biopharmaceuticals and what other biomolecules are in their drug development pipelines that will possibly become the next generation of biopharmaceuticals? In this article I will outline what the current preferences are and look at the myriad of other biomolecules that are potential future biotherapeutics.
Current Biopharmaceuticals
Current biopharmaceuticals can be divided in to two broad areas: therapeutic proteins — probably the most familiar one being insulin– and monoclonal antibodies. Vaccines are also classed as biologics and potentially represent a third arm. Revenue from biopharmaceuticals in 2016 was in the range of $280-300 billion and is forecasted to grow at about 10% for the foreseeable future. One particular biomolecule dominates though in biopharmaceuticals currently and that is the monoclonal antibody.
Following the first commercialisation of a therapeutic monoclonal antibody in 1986, the number approved for use as biopharmaceuticals has steadily increased and in 2016 accounted for five of the top ten selling drugs globally with revenues of $46 billion, as can be seen in this article. This appetite for monoclonal drugs shows no sign of abating with this article on the therapeutic monoclonal antibody market anticipating 2020 revenues of $125 billion.
A recent FiercePharma report also predicted that by 2022 monoclonal antibodies will make up 60% of the top selling cancer drugs. The use of monoclonal antibodies has become so widespread as they offer a multitude of benefits over small molecule drugs such as better targeting, less side effects and well understood modes of action. These benefits and the pipeline of monoclonal antibodies in development ensures that they will be both a current and future biotherapeutic of choice. But other biotherapeutics, some based on monoclonal antibodies and others on different biomolcules, are in development to improve on the current gold standard.
Future Biopharmaceuticals
I say future biopharmaceuticals, but some of these are already being trialled and used; however their use is increasing with the potential to rival monoclonal antibodies in the future. Many of these are however based around monoclonal antibodies.
-
- Antibody-Drug Conjugates (ADCs) – as the name would suggest these are antibody based. A cytotoxic drug is attached to the antibody in order for the antibody to locate and attach to the target cell. The associated drug is then taken in to the cell which is subsequently destroyed by the drug in a highly targeted approach. The antibody can thus be seen as a highly targeted weapons delivery system. Kadcyla is an example of an ADC where the cytotoxic drug entamsine is conjugates to the monoclonal antibody trastuzumab that targets the HER2 receptor on breast cancer cells.
-
- Bispecific Antibodies – again, another antibody-based approach. Here additional specificity is added to an existing antibody so it can bind more than one target; useful for bringing different cells or components together or cross-linking receptors to elicit a response. An example of this approach is blinatumomab, which in 2014 was the first approved bispecific antibody. It binds both the CD3 antigen of a T-cell and the B-cell CD19 antigen so as to bring together a malignant B-cell with a potent cytotoxic T-cell. I recommend this article for a more in-depth look at the bispecific antibody approaches being employed.
-
- Fusion Proteins – this is where proteins are fused together through the joining of two of more genes that encode for separate proteins. This process can occur naturally as with the bcr-abl fusion protein in CML (chronic myeloid leukaemia), or can be genetically engineered to produce biotherapeutics. Typically in biotherapeutics fusion proteins are used to place a human antibody Fc region onto a mouse antibody Fab region so specificity is maintained, but the antibody becomes ‘humanized’ so the body does not see it as foreign and reject it. An example is Enbrel (2016’s third best-selling drug) which consists of a tumour necrosis factor receptor for targeting and an antibody Fc region to stabilise the molecule and improve deliverability.
-
- RNA and DNA Interference and Silencing – most of the attention to date is focused on RNA and RNA interference (RNAi) and RNA antisense technologies. With this approach the RNA molecules prevent gene expression of the target gene(s). The obstacles with this approach are that the RNA-based therapeutics are unstable, hard to deliver and also toxicity is sometimes an issue. However strategies to overcome these are being developed and many such therapies are beginning to enter into the early stages of development.
-
- CAR-T Cell Therapy – chimeric antigen receptors (CARs), alternatively known as artificial T-cell receptors, are genetically engineered receptors that are placed on a T-cell. Typically these engineered receptors offer the specificity of a monoclonal antibody. In CAR-T cell therapy a person’s T-cells are removed and their receptors genetically modified to be specific for an antigen on the target cell. When these T-cells are reintroduced in to the person they then bind their specific target antigen and the T-cell does what a T-cell is designed to do, namely induce apoptosis (programmed cell death) in the target cell.
-
- Biosimilars – you may think that this belongs in the ‘current biotherapeutics’ section, but these are simply a copy of the original biopharmaceutical, so I argue, are a future version of a current therapeutic. They are a similar molecule, addressing the same ailment, but the process to produce such a biomolecule that behaves in a similar way is far from easy.
As you can see there are a variety of different biomolecules and approaches being developed as biopharmaceuticals, however what both current and future biopharmaceuticals have in common is that they are complex molecules that are more often than not produced in a living organism which creates a host of additional problems compared to relatively straightforward chemical synthesis of small molecule drugs. This complexity necessitates thorough characterisation of the biomolecule at all stages from development, manufacture through to formulation.
Biopharmaceutical Characterisation
The complexity of a biopharmaceutical can manifest itself in many ways, but post-translational modifications (PTMs) are the most common. Even relatively minor omissions, additions or pattern changes of PTMs can have a dramatic effect on the biopharmaceutical function, hence the need for complete characterisation to ensure efficacy and safety.
Depending on the challenge, an array of technologies can be used for characterisation, however many workflows contain UHPLC (ultra high-performance liquid chromatography) and mass spectrometry (MS) as these technologies alone or in combination can yield enormous amounts of information on the biomolecule and its PTMs.
Summary
The monoclonal antibody is currently the gold standard biopharmaceutical, but other formats, some of which still rely on the monoclonal antibody as the basis, are in development or have already been commercialised. Whether they will displace the monoclonal antibody in the future, we shall have to wait and see, but indications are that the monoclonal antibody still has a strong future. What binds the current and future biopharmaceuticals together is their complexity and need for thorough characterisation to ensure safety and efficacy. Can your current analytical instruments rise to the challenge this presents?
Additional Resources
-
- Find out more about biopharmaceutical characterisation in our learning centre
-
- Watch this short video on NIBRT’s perspective on biotherapeutic characterisation challenges