Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs using chromatography and mass spectrometry.
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Flame Ionization Detection Crucial for Accurate Blood Alcohol Analysis
By Lori Dolata, 11.09.2015
While watching the local news, you’ve probably heard of people in your area getting arrested for driving under the influence. Across the U.S., driving with a blood alcohol concentration (BAC) (link to chart on CDC website) greater than or equal to 0.08 percent is a crime, with penalties varying by state (link to drunk driving laws by state chart). There is some variation in legal limits in different countries around the world (link to world map with BAC limits).
Measuring BAC
BAC is usually measured as mass per volume. For example, a BAC of 0.03% means there are 0.03 grams of alcohol in 100 grams of a person’s blood. Headspace gas chromatography coupled with a flame ionization detector is one of the most common methods for measuring BAC in forensic labs. This method allows a large number of samples to be analyzed quickly and precisely. Ethanol is the main substance tested for during analysis, but scientists can also look for methanol, acetone, and 2-propanol.
Accuracy is crucial in blood-alcohol analysis, since legal decisions can be made based on a person’s BAC at the time an accident or incident occurs. However, blood is a complex matrix that can have a lot of variation from person to person, for example in the salt or lipid content. So how do forensic scientists and toxicologists make sure their test results are reliable?
Flame Ionization Detector Sensitivity
The typical instrument configuration for analyzing BAC via headspace gas chromatography with flame ionization detection consists of a static headspace instrument for sample introduction, followed by gas chromatography (GC) with two capillary columns for separation, and two flame ionization detectors (FIDs) for detection and quantitation.
Flame ionization detectors are mass sensitive, meaning their measurements are proportional to the mass of the carbon that passes through them. FIDs operate by detecting ions formed during the combustion of organic compounds in a hydrogen flame. The generation of these ions is proportional to the concentration of organic species in the sample gas stream. Hydrocarbons generally have molecular response factors equal to the number of carbon atoms in their molecule. Molecules that contain only carbon and hydrogen respond best to flame ionization detection. As ethanol (link to chemical fact page) is primarily made up of hydrogen and carbon, it can be accurately analyzed by FID.
Tools for Blood Alcohol Content Analysis
There are multiple state-of-the-art tools available for analyzing a large volume of BAC samples quickly and precisely. Gas chromatography systems work well thanks to their versatility, with features like instant-connect modularity. Modules can be installed by the user in less than two minutes, with no special training, dedicated tools or on-site service engineers required. Such modules include the Instant Connect Flame Ionization Detector and the Instant Connect Split Splitless Injector. Another gas chromatography-mass spectrometry system is designed to ensure reliable performance in all ionization modes. Source components are composed of solid, inert material, including a repeller that assists in maintaining instrument performance over time.
Additional Resources
Check out our Forensics Community, a great resource dedicated to our Forensics customers and featuring the latest on-demand webinars, videos, application notes, and more.
    • Rapid, Automated, and Accurate Determination of Volatiles in Human Blood by Headspace Gas Chromatogr… (application note)
Lori Dolata
The Promise of Proteogenomics Precision Medicine
I Love Science Because…
Privacy StatementTerms & ConditionsLocationsSitemap
© 2023 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Go to mobile version