Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs using chromatography and mass spectrometry.
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Sherlock Holmes Brilliant Forensic Scientist
By AnalyteGuru Staff, 03.11.2017
When we think of Sherlock Holmes, Sir Arthur Conan Doyle’s famous detective, we think of a brilliant mind, sharp observation skills and great deductive powers. Actually, he was a highly skilled forensic scientist before the field of forensic science existed. Holmes’ extraordinary talent for deduction is a product of his well-versed training in everything from chemistry to anatomy to ballistics. How did he do it? He integrated his vast knowledge with technology available in the Victorian era, a magnifying glass and a 10-power microscope. Despite a lack of technological options, Holmes was able to decipher the smallest clues and observe the most imperceptible signs to determine the cause of death and figure out who committed the crime. Throughout his cases, Holmes was able to deduce the poison involved in a homicide, typically a heavy metal or natural substance. However, I’m not so sure his forensic prowess alone would work as well in modern day forensic drug testing casework.
While Sherlock Holmes was a fictitious character, modern day forensic scientists use a variety of instruments to identify hundreds of illicit compounds, beyond the typical heavy metals or plant-based poisons used in Victorian England. Technologies such as immunoassays (IA), gas chromatography (GC), liquid chromatography (LC), and mass spectrometry (MS) are routinely used to screen evidence and accurately confirm the identity of the compounds in a sample. Tandem MS (LC-MS/MS) instruments such as the Thermo Scientific„¢TSQ Altis„¢or TSQ Quantis„¢provide confident analysis, quantifying drugs or poisons to below ng/mL concentrations.
As elusive as some of Holmes’ most notorious nemeses, modern criminals have changed their methods with the advancement of technology. With the rise of designer drugs or novel psychoactive substances (NPS) from illegal drug labs, as many as 300 new illicit drugs have flooded the streets in the past 10 years. As soon as an NPS is identified and regulated, a different drug is quickly synthesized, often with closely related properties. Forensic scientists are using high resolution accurate mass spectrometry (HRAM-MS) to further improve their analytical capabilities to identify these fast evolving drugs. Instruments such as the Q Exactive Focus„¢with Orbitrap technology from Thermo Scientific„¢ provide exceptional resolution to 5 decimal places, distinguishing between closely related compounds such synthetic cannabinoids JWH-098 and JWH-081. These two regulated compounds would otherwise be indistinguishable with conventional MS/MS. This resolving power is further put to good use for screening over 1,500 drugs in a single analysis.
Innovations such as mzCloud spectral libraries allow regular updates of newly regulated designer drugs to be shared easily between forensic labs. No doubt modern day forensic scientists have more drugs to look for than Sherlock Holmes did, but mere mortal forensic scientists don’t need the genius of Holmes. By combining the unmatched resolution of Orbitrap technology with the shared data in mzCloud, modern forensic science brings Holmes’ brilliance into the modern laboratory.
Resources:
Download the eBook: Removing Doubt in Forensic Science – Mass Spectrometry Workflows Solving Forensic Cases
Webinar: Novel Psychoactive Substances: Challenges and Choices in Forensic Toxicology
LabManager: The Best Analytical Techniques for Testing Drugs of Abuse
Forensic Learning Center: Learn about forensic analysis methods and technologies
AnalyteGuru Staff
Speciation Using IC ICP MS To Catch a Smooth Criminal
Robustness Enhances Lab Optimization Analysis Processes Part 7: Organic Elemental Analyzers (OEA)
Privacy StatementTerms & ConditionsLocationsSitemap
© 2023 Thermo Fisher Scientific. All Rights Reserved.
Copyright © 2023 · Accelerating Science Multilingual Blogs on Genesis Framework · WordPress · Log in
Talk to us