Advancing PFAS Detection in Pharmaceutical Manufacturing with LC-HRAM-MS
In the realm of pharmaceutical manufacturing, the detection and analysis of perfluoroalkyl substances (PFAS) have become a growing source of concern due to their persistence and potential health risks. The latest application note from SGS presents a groundbreaking approach to PFAS screening and quantification, leveraging the capabilities of the Thermo Scientific Vanquish UHPLC and Orbitrap Exploris 120 high-resolution mass spectrometer (HRAM-MS).
This application note demonstrates the sensitive detection and identification of both known and unknown PFAS compounds in extracts from common pharmaceutical manufacturing and packaging materials. By employing a combined targeted and non-targeted approach, the study highlights how the integration of the Vanquish UHPLC and Orbitrap Exploris 120 HRAM-MS can revolutionize PFAS analysis.
The key benefits of this method include:
  • Combined Targeted and Non-Targeted Screening: One injection allows for both the quantitation of specific PFAS compounds and the screening for additional, previously unidentified PFAS contaminants.
  • Enhanced Analytical Confidence: The use of a PFAS analysis kit and delay column minimizes background PFAS interference, thereby increasing the reliability of the results.
  • Regulatory Compliance: Utilizing Chromeleon CDS ensures that the data acquisition and quantitative analysis comply with 21 CFR Part 11, a crucial requirement for pharmaceutical applications.
  • High Sensitivity and Specificity: The method provides unequivocal identification and quantification of PFAS compounds down to sub-ppb levels.
The study underscores the efficacy of the Orbitrap Exploris 120 HRAM-MS in providing a comprehensive solution for PFAS analysis. By integrating both targeted and non-targeted approaches, this method not only enhances the detection sensitivity and specificity but also expands the scope of detectable PFAS contaminants. This dual approach is a significant advancement in extractables screening, offering pharmaceutical manufacturers a robust tool for ensuring the safety and compliance of their products.
Download our joint application note
Download our joint application note to learn about Comprehensive PFAS screening in pharmaceutical packaging and medical devices by LC-HRAM-MS
This collaboration between SGS and Thermo Fisher Scientific illustrates a leap forward in PFAS screening technology, promising higher quality results, increased confidence, and regulatory compliance in the pharmaceutical manufacturing process.
Jon Bardsley