Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Ion Chromatography / An Easy and Simple Method for Analyzing Common Anions in Drinking Water
An Easy and Simple Method for Analyzing Common Anions in Drinking Water
By AnalyteGuru Staff, 08.17.2020
waterMonitoring of various water sources, such as surface water, groundwater and drinking water, for anions is critical to protecting public health and ensuring a safe and clean environment. As demand for this analysis increases, more labs are taking on this type of testing. Often, monitoring right at the drinking water facility or source water is desired because results will be obtained much faster and treatment processes can be adjusted and modified in a timelier fashion. Having testing capabilities on-site means results are obtained much faster as opposed to having to send samples to a centrally located laboratory, such as one at a public health laboratory. The most common method for measuring anions in drinking water is by ion chromatography.
Ion Chromatography (IC) is the ideal methodology because the technique can separate and quantify all the ions of interest in one analysis. IC also provides the required sensitivity to be able to detect levels at published limits. Also, if other ions or contaminants become important at some point, the analysis parameters can usually be altered to accommodate or add the new ion of interest into the method. For example, many labs will start following a regulated method such as US EPA 300.0, looking at bromide, chloride, fluoride, nitrate, nitrite, phosphate (in the form of Orthophosphate) and sulfate. However, the lab may want to add other ions like bromate, chlorate and chlorite to their testing panel. IC is flexible enough to be able to add these analytes into one analytical method.
For labs who may be unfamiliar with IC, the technique may seem complex and difficult to adopt. Ion chromatography requires some additional considerations as compared to other liquid-based chromatography methods. Most notably, the counterions must be suppressed from the eluent and sample. Counterion suppression also converts the ion of interest to a more conductive form, which boosts the signal, therefore, increasing sensitivity. For more details on suppression, have a look at our in-depth articlehere.
Ion chromatography can be very straightforward and easy, without the need to make large investments. While other technologies use external pumps and columns or chambers, the new Thermo Scientific™ Dionex™ Easion™ Ion Chromatography System makes IC very simple. Only two solutions need to be prepared by dilutions from concentrates and the system equilibrated to be ready to analyze environmental and drinking water samples. Without the need to worry ancillary items, such as auxiliary or peristaltic pumps, IC can be adopted by any lab or testing facility.
To celebrate simplicity in ion chromatography and learn about Thermo Scientific™ Dionex™ Easion™ Ion Chromatography System, download our application note:
1975 text image
Innovations of 1975 and the Birth of the First Commercialized Suppressed Ion Chromatograph
The year 1975 was a remarkable period in the history of tech...
Read More
US EPA Method 1621_1200x600
Understanding PFAS and Combustion-Ion Chromatography
Per- and polyfluoroalkyl substances (PFAS) comprise more tha...
Read More
Robust Analysis of Halogens and Sulfur in Industrial and Environmental Samples
Robust Analysis of Halogens and Sulfur in Industrial and Environmental Samples
In industrial processes, accurately determining halogens and...
Read More
IC Golden Jubilee blog header image
What is the Ion Chromatography Golden Jubilee? Celebrating 50 Years of IC Innovation
The year 2025 marks the golden jubilee of ion chromatography...
Read More
AnalyteGuru Staff
Are You in Control of Your Chromatography Data System
To Query or Not to Query That Is the Question