Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Pharmaceutical / An Introduction to Pharmaceutical Analytical Development
An Introduction to Pharmaceutical Analytical Development
By Aaron Lamb, Vertical Marketing Manager, Pharma and Biopharma, Thermo Fisher Scientific 04.09.2026
What is analytical development?
Analytical development focuses on designing, developing, and validating methods to test and characterize drug substances and products, ensuring accurate measurement of identity, strength, purity, quality, and stability across the product life cycle. It supports formulation, manufacturing, stability studies, and regulatory submissions, helping ensure medicines are safe, effective, and consistently meet regulatory standards.
Scientist using a microscope with test tubes and lab glassware in a research setting
What resources are there for your analytical development needs?
The Thermo Scientific Analytical development brochure covers all aspects of analytical development across the entire pharmaceutical pipeline. Analytical equipment covered in detail are for metabolite identification, extractables and leachables analysis, bioanalysis, genotoxic impurity analysis and quality control batch release testing. An overview of the importance of each and additional key resources are given below.
  1. Metabolite identification
    MetID is a crucial and integral part of drug discovery and development. Confident metabolite annotation generally requires fragmentation data to allow structure elucidation. Rapid metabolism reduces the duration and potential efficacy of pharmaceuticals. Common challenges include limited sensitivity and selectivity in metabolite detection, to learn more about the solutions view this Met ID eBook.
  2. Extractables and leachables analysis
    E&L analysis encompasses characterization and quantification of unknown compounds in pharmaceuticals or medical devices. Comprehensive solutions are available to address the key challenges such as confident identification of unknowns, learn more about addressing these in our E&L solution guide.
  3. Bioanalysis
    Bioanalysis requires the ultimate performance—speed, resolving power, and robustness—all in the presence of a complex biological matrix. With hundreds of samples processed on each system per day, fast separation is required and must remain stable. This in turn means more strain on the resolving power and scan speed of the detector. Challenges include time consuming error prone manual sample preparation, to learn more about how to address these challenges with comprehensive solutions visit the Bioanalysis webpage.
  4. Genotoxic impurity analysis
    To minimize the in-situ formation of nitrosamines for example in pharmaceuticals, either during manufacturing or storage, it is important to accurately assess the precursors such as nitrite and secondary amines as part of risk assessment and QC. To learn more about how to address the common challenges include in situ formation of nitrosamines download our comprehensive Nitrosamine Solution Guide.
  5. Quality control batch release testing
    High throughput and confidence in the results is key in product release. Methods are locked down through validation and need to be transferred across platforms and even laboratories. Common challenges include ensuring 21 CFR part 11 compliant software with mass spectrometry data to learn about how to overcome this challenge with compliance ready software visit this webpage CDS Software—Built for Compliance. Any deviation or performance issues need to be investigated and so robust platforms, and the ability to transfer methods within strict performance criteria, are essential.
To learn more about these topics download the Analytical Development brochure now.
Visit us on LinkedIn: #analyticaldevelopment #pharma #massspectrometry
Automated production line filling and transporting medical injection pens in a factory.
Stability-Indicating HPLC-UV Method for Semaglutide Impurity Analysis
As demand for GLP-1 receptor agonists like semaglutide conti... by Lizzie Gallagher / 03.16.2026
Read More
Illustrated scientists, sustainability icons, and arrow pointing to a laboratory instrument.
Modernizing Routine HPLC Methods: Aligning Productivity, Compliance, and Sustainability
Interested in HPLC method modernization and compliance? I an... by Vincent Lelièvre / 03.03.2026
Read More
pfas image
Understanding PFAS in Pharmaceutical Manufacturing
Introduction to PFAS Per- and polyfluoroalkyl substances (PF... by Aaron Lamb / 08.14.2025
Read More
laptop with ThermoFisher logo
Understanding Nitrosamine Drug Substance-Related Impurities (NDSRIs)
Introduction Nitrosamine drug substance-related impurities (... by Aaron Lamb / 06.26.2025
Read More
Aaron Lamb
Vertical Marketing Manager, Pharma and Biopharma, Thermo Fisher Scientific
What Happens When Glycoproteomics Meets Real-Time LC-MS Decision Making?
When Resolution Changes the Story: Single Quadrupole vs. HRAM GC-MS