Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / GC-MS / Characterizing e-Cigarette Liquid Compounds: Top Questions Answered
Characterizing e-Cigarette Liquid Compounds: Top Questions Answered
By Dominic Roberts, Product Marketing Manager, Orbitrap GC-MS, Thermo Fisher Scientific 03.29.2023
Electronic cigarettes were introduced in 2007 as an alternative to conventional tobacco products, and their use has significantly increased worldwide. Yet despite their growing popularity, little is known about the potential impact of e-cigarettes on human health. This is especially important in relation to the presence of flavoring compounds, solvents, additives and other components intentionally or unintentionally added with unclear long-term effects.
As you seek to understand the opportunities and challenges of analyzing e-cigarette liquid compounds, here are answers to some frequently asked questions:
What are regulatory bodies doing to address the unknown risks of e-cigarettes?
In 2012, the U.S. Food and Drug Administration (FDA) established a list of 93 €œharmful and potentially harmful constituents€ (HPHCs) in cigarette smoke, cigarette filler, and smokeless tobacco products.
Under section 904(a)(3) draft guidance of the Federal Food, Drug and Cosmetic Act (the FD&C Act), only a representative subset of 20 HPHCs to be reported by tobacco product manufacturers for combustible products are detailed.
Additionally, under section 910 draft guidance of the FD&C Act, 29 HPHCs have been outlined in the Premarket Tobacco Products Applications (PMTA) guidance for Electronic Nicotine Delivery Systems (ENDS).
In May 2016 the Tobacco Products Directive (TPD) 2014/14/EU5 introduced new rules for nicotine-containing electronic cigarettes and refill containers (Article 20), to protect human health and meet the obligations of the European Union under the World Health Organization’s WHO Framework Convention on Tobacco Control. In the UK, the majority of the provisions under article 20 are implemented by the Medicines and Healthcare products Regulatory Agency (MHRA). Other EU member states have transposed the EU TPD into their own national laws and assigned competent bodies to oversee.
What analytical technologies are used for the qualitative and quantitative assessment of electronic cigarette liquids (e-liquids)?
The most frequently used technologies to analyze e-liquids are liquid chromatography-mass spectrometry (LC-MS) and gas chromatography-mass spectrometry (GC-MS), but both techniques can have limitations with regard to mass accuracy, sensitivity and linear dynamic range. GC-MS triple quadrupole and GC-FID would typically only be used for quantification of known compounds in e-liquids.
An alternative approach is high-resolution accurate mass (HRAM) mass spectrometry which can achieve confident targeted and non-targeted compound identification and quantitation.
Are there any challenges associated with using these technologies?
There are several analytical challenges associated with the analysis of e-cigarette liquids using GC or GC-MS. To have good coverage of the chemical content, you need to use a GC-MS platform that can sensitively and selectively detect chemical constituents, taking into account the variety and complexity of possible matrices.
GC coupled to high-resolution mass spectrometry is one of the most appropriate, as it offers both the required sensitivity and selectivity. In particular, GC-Orbitrap MS with sub-ppm mass accuracy, versatility for sample introduction, and combined with unique software algorithms for automated deconvolution and extensive spectral libraries, make it a powerful solution for both qualitative and quantitative assessments of e-liquids, all while operating in full scan acquisition mode.
What if I can’t (or don’t want to) use liquid injections?
Although liquid injections are commonly used in GC-MS workflows for this analysis, an alternative is solid phase micro extraction (SPME), which is a solvent-free technique that combines sample extraction with concentration in a single step. It consists of a fused-silica fiber coated with an organic phase that acts by extracting and concentrating the analytes present using selective adsorptive/absorptive processes. The fiber can be exposed to the headspace or via direct immersion in the sample.
Where can I learn more about chemical characterization of e-cigarette liquids?
In a recent study, Thermo Fisher Scientific used specific analytical systems to perform qualitative analysis of e-liquids on a targeted subset of the US FDA list of HPHCs. The study also used an untargeted approach to screen the samples for other potential toxic chemicals that may be present. The study demonstrated that using specific GC-MS technology, with unique intuitive software workflows for automated deconvolution and extensive spectral libraries, it’s possible to achieve excellent analytical results for e-liquids.
Read more about the study and workflow.
Read the Knowledgebase post: How to characterize chemicals in e-cigarette liquids with Orbitrap GC-MS
We encourage you to subscribe to the AnalyteGuru blog to stay informed when new topics on characterizing e-cigarette liquids are released.
Gloved hands holding a blood sample swab and vial with chemical structures overlaid.
Sharper, Faster, Smarter: HRAM Orbitrap Mass Spectrometry Meets Postmortem Toxicology
In forensic toxicology, understanding what is in a sample an...
Read More
Ocean to the laboratory; story of microplastics
From the Ocean to the Laboratory: How Scalable Pyrolysis-GC/MS Reveals the Hidden Story of Microplastics
Plastics have become part of everything we touch — from th...
Read More
Sheffield teaching hospital NHS foundation
Beyond Blood: Sheffield Teaching Hospitals NHS Foundation Trust Develops HRAM-Powered Method for Post-Mortem Toxicology Analysis
In a recent study published in the Journal of Analytical Tox...
Read More
Pesticides
Pesticide Residue Analysis in the 21st Century: Innovations in Sample Preparation and Detection Using the Thermo Scientific TSQ 9610 GC-MS/MS
Introduction The accurate detection of pesticide residues in...
Read More
Dominic Roberts
Dominic Roberts has been closely involved with the Thermo Scientific GC Orbitrap MS since its launch in 2015. He started at Thermo Fisher Scientific as a Senior Applications Specialist for the product and is currently the GC Orbitrap MS Product Marketing Manager. Dominic is an experienced analytical chemist and mass spectrometrist with more than 20 years of experience using state-of-the-art analytical systems. He has worked in both a regulatory and industrial environment with GC–MS and LC–MS systems to investigate a variety of chemicals in food, biological, and environmental samples.
Pioneering Proteomics ‘Mousetraps’: Webcasts Showcase Cutting-Edge Researchers
ACS Award 2023 in Chromatography Presented to Christopher Pohl