Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Combustion Ion Chromatography / Confident Total Organic Fluorine Analysis in Textiles with the Cindion Combustion-Ion Chromatography System
Confident Total Organic Fluorine Analysis in Textiles with the Cindion Combustion-Ion Chromatography System
By Jingli Hu, Application Staff Scientist, Thermo Fisher Scientific 01.22.2026
Per- and polyfluoroalkyl substances (PFAS) are widely used in textile products to give them water-repellent and stain-resistant qualities. As regulations limit PFAS in consumer products, manufacturers and testing laboratories need analytical tools that can reliably detect fluorinated compounds, even when PFAS structures are unknown.
PFAS in textiles
Our recent application note, Screening of PFAS in textiles: Utilizing a new combustion IC system for total organic fluorine (TOF) analysis, highlights how the Thermo Scientific Cindion Combustion-Ion Chromatography (C-IC) System provides a streamlined, sensitive, and robust approach to measuring TOF in textile materials. TOF analysis provides a comprehensive way to evaluate PFAS presence by quantifying all organically bound fluorine in a sample, regardless of chemical identity. This is particularly valuable for textiles, where PFAS may be embedded within fibers and are not always easily extractable. Targeted LC-MS methods typically measure only extractable PFAS, whereas combustion ion chromatography captures all organically bound fluorine, including non-extractable fluorinated species. For textiles, where PFAS may be embedded in fibers and not easily extractable, TOF offers a more complete representation than targeted methods.
The Cindion platform simplifies TOF analysis by combining combustion, absorption, and ion chromatography into a unified, software-controlled workflow. Across six textile types (canvas, waterproof fabrics, and tablecloth materials), the Cindion C-IC system delivered TOF values that were between 100% and 105% compared with those obtained from a previously used combustion IC platform. The relative standard deviation for each sample was also comparable at <8%. This demonstrates strong accuracy and reproducibility, essential for compliance-focused workflows.
As PFAS regulations broaden into consumer goods, laboratories must adopt methods that are:
  • Sensitive enough to detect trace fluorine
  • Reliable across diverse, difficult-to-extract materials
  • Fast and automated to support higher-throughput testing
  • Comprehensive enough to capture all organically bound fluorine, regardless of extractability
The Cindion C-IC System meets all of these requirements. By directly combusting textile samples without an extraction step, it provides a streamlined workflow with low background contamination for determining fluorine levels in textiles. This TOF method delivers greater confidence as industries work to comply with evolving regulations.
Visit the Cindion combustion ion chromatography system page to learn how the Cindion C-IC system can elevate your analytical capabilities and ensure regulatory compliance.
Visit us on LinkedIn: #PFAS #CombustionIC #CIC #Cindion
pfas image
Understanding PFAS in Pharmaceutical Manufacturing
Introduction to PFAS Per- and polyfluoroalkyl substances (PF...
Read More
Food packaging
Enhancing PFAS Analysis in Food Contact Materials Using Combustion Ion Chromatography
Per- and polyfluoroalkyl substances (PFAS) have been used in...
Read More
US EPA Method 1621_1200x600
Understanding PFAS and Combustion-Ion Chromatography
Per- and polyfluoroalkyl substances (PFAS) comprise more tha...
Read More
Robust Analysis of Halogens and Sulfur in Industrial and Environmental Samples
Robust Analysis of Halogens and Sulfur in Industrial and Environmental Samples
In industrial processes, accurately determining halogens and...
Read More
Jingli Hu
Jingli Hu is a Senior Applications Chemist at Thermo Fisher Scientific in Sunnyvale, CA. She specializes in ion chromatography (IC) applications, focusing on food, environmental, and biopharma markets.
How to Cut HPLC Sample Preparation Time for Derivatizing Amino Acids
Sharper, Faster, Smarter: HRAM Orbitrap Mass Spectrometry Meets Postmortem Toxicology