This month the European Medicines Agency (EMA) issued further revisions to the ever-changing guidance for drug producers when it comes to nitrosamine testing. In 2018 the Federal Drug Administration (FDA) showed the presence of n-nitroso dimethylamine (NDMA) at unacceptable levels in angiotensin receptor blockers (ARBs), and since then further nitrosamine compounds were found in other drug products such as metformin. This topic has had marketing authorization holders (MAHs) around the globe scrambling to better understand the impact on their products, implement risk assessments, and devising analytical strategies.
Further background on nitrosamine testing strategies can be found here within a handy toolkit.
The recent announcement by the EMA has three highlights;
- Introduction of NDPA as a new nitrosamine of interest
- Practical options for deciding on limits when multiple nitrosamines are present
- Multi-step review of the manufacturing process
But what does this mean to my analytical methods and testing strategies?
Advanced LC-MS solutions with increased sensitivity and separation efficiency for nitrosamine detection
It is now more important than ever to stay ahead of the changes. Implementing proven instruments and solutions with increased sensitivity and separation efficiency to detect a wide range of nitrosamine impurities, including the new impurity NDPA, to help future-proof your methods. This ensures your routine monitoring can accurately and reliably meet worldwide regulatory requirements.
Application benefits:
- Detection and quantification of 11 nitrosamines in a single liquid chromatography, high-resolution accurate-mass mass spectrometry (LC-HRAM-MS) method
- Quantitation of nitrosamine impurities in metformin drug products below the daily acceptable intake level, that meets U.S. FDA regulatory guidelines
- Compliant data acquisition, processing and auditing built for a U.S. FDA 21 CFR part 11 compliant environment
Advanced Ion Chromatography solutions to accurately and quickly monitor nitrosamine precursors
Proactively monitoring drug products and ingredients can help you understand the risk level and impact when the presence of nitrosamine precursors is suspected to be present. Predictions can be made through understanding the chemical properties of each stage of the production process, but we also need to be aware of potential contaminated ingredients.
Selecting the right technologies to enable the analysis and monitoring of nitrosamine precursors — such as nitrite — ensures you know exactly what is in the raw ingredients. This helps to understand the risk of nitrosamine formation in the manufacturing process and product storage.
Application benefits:
- Cost-effective determination of nitrite in pharmaceuticals by coupling IC with UV absorbance detection
- The method is accurate and precise due to the high reproducibility of the reagent-Free ion chromatography system
- This method is applicable to the determination of nitrite throughout the manufacturing process of a drug product to assess the likelihood of nitrosamine formation
Receive updates
on nitrosamine
analysis and other
Pharma & Biopharma
topics of interest
Exploring a Versatile LC–HRMS Strategy for NDSRI Determination in Beta Blocker Drug Substances
The increasing focus on nitrosamine impurities has prompted ...
Read More
Confident Total Organic Fluorine Analysis in Textiles with the Cindion Combustion-Ion Chromatography System
Per- and polyfluoroalkyl substances (PFAS) are widely used i...
Read More
Understanding Nitrosamine Drug Substance-Related Impurities (NDSRIs)
Introduction Nitrosamine drug substance-related impurities (...
Read More
What is the Ion Chromatography Golden Jubilee? Celebrating 50 Years of IC Innovation
The year 2025 marks the golden jubilee of ion chromatography...
Read More