Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Environmental / Hexane in Food: When Processing Chemistry Meets Public Health
Hexane in Food: When Processing Chemistry Meets Public Health
By Petra Gerhards, Regional Marketing Manager GC and GC/MS, EMEA, Thermo Fisher Scientific 02.04.2026
Hexane is one of the most widely used extraction solvents in the global food system. Derived from petroleum, it plays a central role in efficiently extracting vegetable oils from crops such as soy, sunflower, rapeseed, and corn. The same process produces protein-rich oilseed meals that are widely used in animal feed. From a technological perspective, hexane is effective, inexpensive, and well understood. From a toxicological and regulatory perspective, however, it is increasingly under scrutiny.
Curl of butter on a butter knife resting on a dark surface.
In the European Union, “technical hexane” is authorized as a food extraction solvent under Directive 2009/32/EC. Its last full safety assessment by the EU’s Scientific Committee on Food dates back to 1996. In 2024, the European Food Safety Authority (EFSA) concluded that this assessment is no longer sufficient and formally called for a re-evaluation. The reasons are scientific rather than sensational: technical hexane is not a single pure compound, its impurities are poorly specified, human absorption of n-hexane is well documented, and exposure—particularly for infants and young children—may have been underestimated under current regulatory assumptions.
At the same time, public attention has intensified. Investigations commissioned by Greenpeace reported detectable hexane residues in a wide range of everyday foods, including vegetable oils, butter, milk, infant formula, and poultry. While the measured levels generally fall within current legal limits, these findings highlight a critical gap between regulatory compliance and public expectations of food safety and transparency. Processing aids like hexane are not listed on labels, even though trace residues may remain in final products.
From a scientific standpoint, the debate raises several important questions. How robust are existing toxicological datasets for chronic, low-level exposure? Do current analytical methods sufficiently characterize impurities and residues across complex food matrices? And should efficiency-driven processing choices be reconsidered when safer—though potentially less economical—alternatives exist?
EFSA’s conclusion is cautious but clear: there is no immediate acute health concern, but the available evidence is insufficient to confidently assess long-term risks. As new toxicological data emerge and analytical capabilities improve, hexane has become a case study in how legacy food technologies can outpace the science and regulation designed to govern them. The outcome of the ongoing re-evaluation may shape not only future solvent use, but also broader expectations around transparency, precaution, and trust in the food system.
Global inconsistencies in safety standards
Hexane regulation varies widely across jurisdictions. European limits are among the most conservative, yet they rely heavily on industry-generated data. In other regions, allowable residue levels are significantly higher, and in some cases, hexane use in food and feed is largely unrestricted. This variability complicates global risk assessment and raises questions about harmonization and scientific consistency in food safety governance.
Beyond compliance: Rethinking food processing choices
The ongoing re-evaluation of hexane illustrates a broader challenge in food science: technologies adopted for efficiency and cost-effectiveness may persist long after their safety assumptions become outdated. The issue is not whether hexane poses an immediate public health crisis—EFSA has explicitly stated that it does not—but whether current scientific evidence is sufficient to justify its continued widespread use without further scrutiny.
Advances in analytical chemistry now allow more sensitive detection of residues and impurities, while modern toxicology places greater emphasis on long-term, low-dose effects. At the same time, alternative extraction technologies, though less efficient, are becoming more viable as sustainability and safety considerations gain weight.
What are the best methods for detection?
Detection options
  • HS-GC-FID: robust, sensitive for hydrocarbons, widely used for routine control (often aligned with “official” oil methods such as AOCS workflows referenced in the literature).
  • HS-GC-MS: adds spectral ID confirmation (useful when there are interferences or you want stronger identity evidence).
Quantification
  • Prefer internal standard calibration (often a similar volatile hydrocarbon not expected in the sample) and matrix-matched calibration where feasible.
Conclusions
EFSA concluded that there is a need to re-evaluate the safety of technical hexane used as an extraction solvent in the production of foodstuffs and food ingredients. A key aspect of this re-evaluation concerns the accurate characterization and quantification of hexane residues and related hydrocarbons, where gas chromatography (GC) plays a central role. Robust GC-based analytical methods are essential to reliably determine residual solvent levels, distinguish between different hexane isomers and impurities, and ensure compliance with regulatory limits.
Advances in GC methodology, including improved separation, sensitivity, and detection capabilities, can significantly reduce analytical uncertainty and support more precise exposure assessments. High-quality GC data are therefore critical for refining dietary exposure estimates, identifying potential sources of variability, and supporting a science-based risk assessment. Strengthening analytical controls through validated GC methods will be an important component of addressing EFSA’s safety concerns and ensuring continued consumer protection.
Visit us on LinkedIn: #hexaneSafety, #GC-FID #GC-MS
MOSH MOAH image
MOSH/MOAH Analysis: Technology, Regulation, and the Road Ahead
Mineral oil hydrocarbons (MOSH) and mineral oil aromatic hyd...
Read More
wine bottle image
New European Wine Scandal on PFAS in Wine
Introduction PFAS, also known as “eternity chemicals&#...
Read More
Pesticides
Pesticide Residue Analysis in the 21st Century: Innovations in Sample Preparation and Detection Using the Thermo Scientific TSQ 9610 GC-MS/MS
Introduction The accurate detection of pesticide residues in...
Read More
Dr Kerstin Krätschmer
GC Orbitrap Guru Series: Dr. Kerstin Krätschmer
It’s been 10 years! Yes, in 2015 “gas chromatography...
Read More
Petra Gerhards
Petra Gerhards, Dipl-Ing, is Regional Marketing Manager of GC and GC-MS for EMEA at Thermo Fisher Scientific. She has more than 29 years of experience in the fields of GC-MS, SPE and LC-MS. Since joining the regional team she has contributed to workflow solutions combining vials and closures with SPE solutions, GC-MS and LC-MS. She works with KOL's on data for regional specific marketing campaigns, organizes in-house seminars and works on customer specific solutions. Her main expertise is in the field of doping and drugs-of-abuse analysis.
Sharper, Faster, Smarter: HRAM Orbitrap Mass Spectrometry Meets Postmortem Toxicology
From Scattered Files to Clear Insights (Part 1): When Lab Data Review Feels Like Detective Work