Figure 1 below shows results from the analysis of a 50% sodium hydroxide sample produced from a diaphram cell process that are within trace contaminant specifications for chlor-alkali producers.
Figure 2 below depicts the two-valve configuration for single-pass neutralization with either one of our high-pressure ion chromatography systems (Thermo Scientific Dionex ICS-5000+ Reagent-Free HPIC system) or one of our integrated ion chromatography systems (Thermo Scientific Dionex ICS 2100 Integrated IC System with Electrolytic Eluent Generation and Sample Preparation).
Figure 3 below depicts the configuration for double-pass neutralization with either an integrated or a high-pressure ion chromatography system (links to systems above). Here, the caustic sample is first loaded into a 25μL sample loop. The sample is then switched in line and the concentrated base from the sample loop is directed to the self-regenerating neutralizer (Thermo Scientific Dionex SRN Self-Regenerating Neutralizer), where the sample is partially neutralized, then transferred to the 5 mL loop. The recycle valve is then actuated to direct the sample through the neutralizer a second time to complete the neutralization. Following the double-pass neutralization process, the sample anions can be focused onto a concentrator column before eluting them onto an analytical column.
The single-pass neutralization pretreatment with the new neutralizer provides a more efficient alternative to the double-pass neutralization for the analysis of anions in concentrated caustics. Results by the single pass method with ion chromatography are shown to satisfy purity objectives in chlor-alkali processes by measuring trace levels of chlorate, chloride, and sulfate in a faster more effective manner.
Sample Preparation Conditions
Sample | Diaphram Caustic 1:500 dilution with DI water |
Eluent | DI Water with Anion Trap Column (9×75 mm) |
Flow Rate | 1 mL/min |
Injection Volume | 25 μL |
Temperature | 30 0C |
Neutralization | Dionex AERS 500, 4 mm Suppressor (P/N 082542) |
Concentrator | UTAC-XLP1 Anion Concentrator Column |
Analytical Conditions
Column | Thermo Scientific Dionex IonPac AS19 (2×250 mm) |
Eluent | 10 mM KOH 0-12 min, 10-45 mM KOH 12-25 min |
Eluent Source | Thermo Scientific Dionex EGC 500 KOH Cartridge |
Flow Rate | 0.5 mL/min |
Temperature | 30 0C |
Detection | Suppressed conductivity Dionex AERS 500, 2 mm Suppressor (P/N 082541) AutoSuppression, recyle mode |
Additional Resources
Do check out our Chemical Analysis and Manufacturing web page which is an informative resource dedicated to our Chemicals and Polymers customers and features the latest on-demand webinars, videos, application notes, and more.
Is chlor-alkali raw material testing, process analysis, and QA by IC, HPLC, and ICP-OES of interest to your laboratory? If so, I would like to hear your thoughts and experiences.
/* Style Definitions */ table.MsoNormalTable {mso-style-name:”Table Normal”; mso-tstyle-rowband-size:0; mso-tstyle-colband-size:0; mso-style-noshow:yes; mso-style-priority:99; mso-style-qformat:yes; mso-style-parent:””; mso-padding-alt:0in 5.4pt 0in 5.4pt; mso-para-margin-top:0in; mso-para-margin-right:0in; mso-para-margin-bottom:10.0pt; mso-para-margin-left:0in; line-height:115%; mso-pagination:widow-orphan; font-size:11.0pt; font-family:”Calibri”,”sans-serif”; mso-ascii-font-family:Calibri; mso-ascii-theme-font:minor-latin; mso-fareast-font-family:”Times New Roman”; mso-fareast-theme-font:minor-fareast; mso-hansi-font-family:Calibri; mso-hansi-theme-font:minor-latin;}