Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Pharmaceutical / Overcoming the Challenges of Nitrosamine Analysis
Overcoming the Challenges of Nitrosamine Analysis
By AnalyteGuru Staff, 08.21.2020
With roughly 50% of the world population taking prescription drugs, it is important to ensure that pharmaceutical products are safe. Nitrosamine impurities have been widely publicized recently. A year ago we reported the “Valsartan crisis” which detailed the recall. In fact, nitrosamines have been found in different types of angiotensin II receptor blocker (ARB) medicines, and more recently in histamine-2 receptor blocker medicines. It is essential to analyze nitrosamines that can unintentionally occur in pharmaceutical products as they are carcinogenic impurities. The regulatory requirements demand high sensitivity to meet the low reporting limits. The analytical instrumentation must not only be sensitive but also selective to avoid false-positive non-compliant results. The stringent limits imposed by the current regulations will be lowered at the end of the interim period (April 2021), increasing the challenges for analytical science laboratories.
The U.S. Food and Drug Administration (FDA), the European Medicines Agency (EMA) and other regulatory bodies have published several analytical methods that may be considered when determining nitrosamine content in the active pharmaceutical ingredient (API) or finished pharmaceutical product (FPP). At Thermo Fisher, we recently consolidated the methods for GC-MS analysis of nitrosamines in pharmaceutical products into a single case study. This study was published in collaboration with pharmaceutical testing labs like Laurus and SGS and looks at GC-MS and GC-MS/MS coupled with both liquid injection and headspace sampling for quantitative analysis according to the published USFDA methods and high-resolution accurate mass. The latter providing the added advantage of high selectivity and accurate mass confirmation of nitrosamines, and furthermore offers an expanded scope of analysis including options for retrospective examination of samples.
To know more, explore the dedicated Thermo Scientific web page on Nitrosamine Impurities Analysis and discover additional resources:
    • Webinar
    • App note
    • Journal article
    • Nitrosamine impurity analysis web page
On LinkedIn? Visit our LinkedIn page #nitrosamines #pharmaceutical
Automated production line filling and transporting medical injection pens in a factory.
Stability-Indicating HPLC-UV Method for Semaglutide Impurity Analysis
As demand for GLP-1 receptor agonists like semaglutide conti...
Read More
Colorful capsules and pills scattered beside an open prescription bottle.
Exploring a Versatile LC–HRMS Strategy for NDSRI Determination in Beta Blocker Drug Substances
The increasing focus on nitrosamine impurities has prompted ...
Read More
pfas image
Understanding PFAS in Pharmaceutical Manufacturing
Introduction to PFAS Per- and polyfluoroalkyl substances (PF...
Read More
laptop with ThermoFisher logo
Understanding Nitrosamine Drug Substance-Related Impurities (NDSRIs)
Introduction Nitrosamine drug substance-related impurities (...
Read More
AnalyteGuru Staff
To Query or Not to Query That Is the Question
What Is Automated Wet Chemical Analysis?