Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Sample Preparation / Sample Preparation in Food Analysis: Why is it Such a Headache?
Sample Preparation in Food Analysis: Why is it Such a Headache?
By Petra Gerhards, Regional Marketing Manager GC and GC/MS, EMEA, Thermo Fisher Scientific 06.18.2021
Sample matrix
There are as many sample preparation options available as there are matrices. Some are more regulated than others, but it all starts by looking at the matrix under consideration.
A highly water-based matrix is a different challenge than a fat-rich or protein-containing matrix. The job of sample preparation is to separate the analytes from the matrix and then concentrate the analytes to make them analytically compatible so the required detection limits can be reached.
Sample prep in food analysis
Think about the nastiest matrix you can. For some, this is a meat sample containing maggots. For others, it might be rotten fish. However, this is not what makes it complicated. The matrix itself, as well as the analytes and their chemical properties, are the base for the right sample preparation.
A lot of errors occur during sample collection, storage, and transportation. There are a lot of possibilities where things can go wrong and where contamination can occur.
Most samples, if not all, must be homogenized so that a representative sample can be taken for analysis. If this is not done in the right environment, for example, using cooled ball mills to prevent heat generation and thus degradation, that’s where the problem starts. You do not want to lose analytes during the whole process. That is critical for the result. For that reason, there are a lot of parameters that need to be taken into consideration depending on what you are looking for.
Sample preparation
If you are looking into solids or oil, liquid-liquid extraction (LLE) is the best first route. You need to get the particles (e.g., matrix) removed and extract the analytes into a more analytically compatible solution. Now, this is universal enough, but is it clean enough?
The analytical system can be easily contaminated and requires a lot of maintenance afterward, thus cleaning becomes necessary. That costs time and resources, not what’s needed in a lab that needs to produce results.
That’s where special cleanups such as solid-phase extraction (SPE) come into place. Sure, this is more costly and labor-intensive, however, it gives you more specific and analytically compatible results. The more specific you can get, the better the detection limits will be. There are a lot of different options and it all depends on the chemistry and properties of your analytes. There are solutions, proven and widely used.
Fig. 1 shows the objectives of sample preparation to clean, concentrate, and the analytical combability.
Sample prep objectives
Fig. 1 Objectives of sample preparation.
Another route might be QuEChERS, which is a solid-liquid extraction (SLE) that may also require an SPE step afterward.
If you have a liquid sample you might dilute, derivatize, and inject, how sure are you that the matrix doesn’t contaminate your analytical system? The better the cleanup is, the better the results.
If the focus is on volatile substances, there are special methods to treat the sample as well. That might be static headspace (HSS), solid-phase micro-extraction (SPME), or, in some cases, purge and trap (P+T).
An additional option is accelerated-solvent extraction (ASE). This technology is especially useful for difficult matrices such as honey or Dioxin-containing samples.
Sometimes it can be necessary to adopt methods, existing or new, to a group or one specific new analyte that was not on the radar screen two years ago. The same strategy applies — matrix and analyte properties determine the sample preparation.
It is always the goal to find a fast, cheap, and reliable method. However, sometimes a little bit more effort is recommended to really accomplish the necessary detection limits, and infant food is a completely different story when it comes to detection limits and sample prep.
Conclusion
Always know what you want to get rid of and what you need to detect, and how low you can go. That is the bottom line for food analysis and sample preparation. It is a good starting point for our experts to find the optimal solution for your analytical challenge.
Helpful links
QuEChERS and ASE:
https://analyteguru.com/webinars/part-1-of-4-tips-and-tricks-for-sample-preparation-using-quechers-a…
QuEChERS:
https://analyteguru.com/resources/the-quechers-three-minutes-experience-screening-targeted-data-anal…
QuEChERS product information: https://www.thermofisher.com/search/browse/featured/de/de/80013164/Quechers%20Dispersive%20Spe
SPE extraction guide:
https://www.thermofisher.com/de/de/home/industrial/chromatography/chromatography-sample-preparation/…
SPE consumables:
https://www.thermofisher.com/de/de/home/industrial/chromatography/chromatography-sample-preparation/…
Chromatography consumables now available online:
https://www.thermofisher.com/de/de/home/products-and-services/promotions/industrial/chromatography-c…
Two scientists in lab coats reviewing analytical data on a computer beside lab instrumentation.
Internal Standards in ICP-OES: A Practical Guide to Better Accuracy and Precision
In ICP-OES analysis, maintaining reliable accuracy and preci...
Read More
Assorted bottles of cooking oils with olives and herbs on a wooden surface.
Ask the Experts: Practical Answers to Today’s Biggest MOSH/MOAH Analysis Questions
Introduction MOSH and MOAH analysis remains one of the most ...
Read More
Sample Preparation Strategies Demystified: Choosing the Right Fit for Your Toxicology Workflow
Smarter sample preparation for LC-MS: from simplicity to sel...
Read More
Pesticides
Pesticide Residue Analysis in the 21st Century: Innovations in Sample Preparation and Detection Using the Thermo Scientific TSQ 9610 GC-MS/MS
Introduction The accurate detection of pesticide residues in...
Read More
Petra Gerhards
Petra Gerhards, Dipl-Ing, is Regional Marketing Manager of GC and GC-MS for EMEA at Thermo Fisher Scientific. She has more than 29 years of experience in the fields of GC-MS, SPE and LC-MS. Since joining the regional team she has contributed to workflow solutions combining vials and closures with SPE solutions, GC-MS and LC-MS. She works with KOL's on data for regional specific marketing campaigns, organizes in-house seminars and works on customer specific solutions. Her main expertise is in the field of doping and drugs-of-abuse analysis.
Combining HPAE with Orbitrap MS for Improved Glycoprotein Characterization
Charged Aerosol Detection (CAD): Inspiring Next-Generation Separations