Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Food and Beverage / Simultaneous UHPLC Analysis of Water- & Fat-soluble Vitamins With Dual LC
Simultaneous UHPLC Analysis of Water- & Fat-soluble Vitamins With Dual LC
By Dr. Kimberly M. Lincoln, HPLC, Thermo Fisher Scientific 02.22.2023
There’s no doubt we’re all much more health conscious since COVID-19 came into our lives (like a wrecking ball), spurring a surge in healthier lifestyles and diets. One example is the supplement market boom.
Recent data published by Grand View Research projects the vitamin supplement market to $71 billion in global revenue by 2028. Another insightful takeaway from this report is around 40 percent of consumers opt for a multivitamin product versus a single vitamin.
Needless to say, there’s A LOT of money to be made in the multivitamin business. And a huge cash market is likely to attract professional fraudsters that sell adulterated products, which is why analytical testing to ensure product quality and purity is critical for consumer trust and safety.
But analytical testing of multiple vitamins using a single method is challenging due to the opposite polarities of different vitamins.
There are ways to overcome these limitations by using switching valves or delayed injections, but the disadvantages include:
  • Cumbersome CDS programming
  • Complex LC hardware setup
  • Low analytical throughput
  • Poor resolution
This is where the benefits of having a Dual LC system come into play.
Benefits of Dual LC instruments
You can think of Dual LC instruments as twins – fraternal or identical – depending on what your analysis entails. This multichannel system has two pumps, two columns, two injection ports, and two detectors and enables you to run two methods independently, at the same time (Figure 1).
Figure 1. Fluidic setup of Thermo Scientific Vanquish Flex Duo System for Dual LC.
The major perks are:
  • No extra bench space or head count for operation
  • 100 percent increase in analytical throughput
  • Better-optimized methods
  • Simple to implement
Even better, this modern technology allows you to eliminate problems like the precipitation of buffers in high-organic mobile phases resulting from solvent incompatibility in the shared flow path.
A simple method for multivitamin analysis with Thermo Scientific™ Vanquish™ Duo UHPLC Systems for Dual LC
One beneficial application of Dual LC applies to the analysis of vitamins, in particular multivitamin supplements (Figure 2). As described in our application note, Dual LC enables you to of two different columns and methods to analyze vitamins of opposite polarity at the same time.
Figure 2. Separation of a water-soluble vitamin standard (left) and fat-soluble vitamin standard (right) on an Acclaim PA2 column (150 × 2.1 mm, 2.2 µm) recorded at 210 nm and 280 nm, respectively.
Scale your productivity with our Dual LC solutions
Looking for a way to scale your analytical productivity but don’t have the bench space or budget for more headcount? Watch this video (See productivity in a new light) to learn how our Dual LC instruments can boost your analytical throughput by 100 percent versus a single channel system.
P.S. Don’t forget to follow our LinkedIn page to stay in the know with all things #VanquishDuo.
Pre-configured LC_MS workflow solutions
Maximize Performance with Specialized LC-MS Workflows — and Save Up to 40%!
When it comes to clinical research, precision, reliability, ...
Read More
MOSH MOAH image
MOSH/MOAH Analysis: Technology, Regulation, and the Road Ahead
Mineral oil hydrocarbons (MOSH) and mineral oil aromatic hyd...
Read More
Automated LC-MS Sample Cleanup Using Turbulent Flow Chromatography
Too much interfering matrix? Discover how to streamline your...
Read More
Advancing PFAS Detection in Pharmaceutical Manufacturing with LC-HRAM-MS
Advancing PFAS Detection in Pharmaceutical Manufacturing with LC-HRAM-MS
In the realm of pharmaceutical manufacturing, the detection ...
Read More
Dr. Kimberly M. Lincoln
Dr. Kimberly M. Lincoln is a marketing specialist in the HPLC division at Thermo Fisher Scientific. She holds a Ph.D. in bioinorganic chemistry and a B.S. in chemistry, with professional experience in digital marketing, medical device manufacturing, and chemical synthesis. Kimberly enjoys writing educational chemical content as a guide to help customers solve analytical problems. She is passionate about living a clean lifestyle and keeping the planet healthy for future generations. In her spare time, Kimberly loves to cook, cycle, attend rock concerts, and spend time with her pit bulls.
Turnkey Wet Chemistry Analysis: Top 7 Questions Answered
Let’s Have the Talk, the Procurement Talk