Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Food and Beverage / Acrylamide in Coffee? Use This Robust GC-MS Method to Analyze It
Acrylamide in Coffee? Use This Robust GC-MS Method to Analyze It
By Adam Ladak, Global Product Marketing Manager for GC quadrupole MS 02.28.2020
Mornings in the dead of winter are hard to deal with. The sun has not risen, it’s below freezing, and the only thing that gets me going is a nice cup of coffee. Some would say I am a coffee snob as I grind my own beans and use a French press to brew up the delicious black gold. However, the caffeinated lifeblood that puts a spring in our steps in the morning could have an ingredient that is not listed on the packaging.
coffeeAcrylamide (2-propeneamide) is a chemical that has been found in certain cooked foods including roasted coffee. Acrylamide is highly toxic, can cause neurotoxicity, genotoxicity, and reproductive harm, and is a likely human carcinogen. Due to the toxicity of the compound, the food agencies around the world have introduced guidance for the monitoring of Acrylamide in various foodstuffs.
The traditional method of analyzing acrylamide in food is using LC-MS/MS but requires labor-intensive complex extraction techniques which are time-consuming and use large amounts of organic solvents. GC-MS has also been used but traditionally uses derivatization by bromination which takes long periods of time and can be unstable during the analysis.
In order to overcome the challenges in this analysis, Thermo Fisher Scientific scientist Jane Cooper has developed a new analytical method. The work includes the use of the ISQ™ 7000 Single Quadrupole GC-MS and a simple acetonitrile extraction followed by silylation derivatization. This method has been proven to be robust, simple and cost-effective. Using this analytical method, we can be sure that the coffee we so desperately need in the morning is safe for consumption with no unexpected side effects. For more information on the full analytical method read the full application note here.
[A] Robustness data shown as consistent peak area counts for acrylamide determined in QC ground coffee samples spiked at 200 ppb (equivalent to 1000 µg/kg), analyzed mid (inj. no. 46–48), and end (inj. no. 79–92) of a 99-injection analytical sequence, containing derivatized blanks, calibration standards, crisp, instant coffee, and ground coffee samples. [B] overlaid EIC (m/z 128) of the QC ground coffee sample (n=16 injections) analyzed across the whole analytical sequence. For all QC ground coffee samples containing acrylamide at the 200 ppb level across the analytical sequence of 99 injections, the calculated %RSD absolute peak area counts was 2.9.
Ocean to the laboratory; story of microplastics
From the Ocean to the Laboratory: How Scalable Pyrolysis-GC/MS Reveals the Hidden Story of Microplastics
Plastics have become part of everything we touch — from th... by Dr. Daniela Cavagnino / 12.11.2025
Read More
wine bottle image
New European Wine Scandal on PFAS in Wine
Introduction PFAS, also known as “eternity chemicals&#... by Petra Gerhards / 06.27.2025
Read More
Pesticides
Pesticide Residue Analysis in the 21st Century: Innovations in Sample Preparation and Detection Using the Thermo Scientific TSQ 9610 GC-MS/MS
Introduction The accurate detection of pesticide residues in... by Petra Gerhards / 06.02.2025
Read More
Dr Araceli Rivera-Pérez
GC Orbitrap Guru Series #3: Dr Araceli Rivera-Pérez
Hey everyone! The response to my second blog with Brooklynn ... by Dominic Roberts / 06.02.2025
Read More
Adam Ladak
Adam Ladak is the global product marketing manager for GC quadrupole mass spectrometry at Thermo Fisher, based in the UK. He started his career working in a contracting testing lab analyzing sports supplements and horse feed for prohibited substances. He has over 20 years of experience in the analytical testing industry and has worked for leading instrument manufacturers in various roles from an application specialist to a global marketing role focused on food and environmental testing. Adam is passionate about utilizing our GCMS instrumentation to meet the customer needs and solve real world problems.
Top 3 Scan Modes for Small Molecule Quantitation Using Mass Spectrometry
Accelerate Analysis of Trace Elements for Routine Laboratory