Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Isotope Ratio MS / Using Isotope Analysis to Understand Phosphorus Cycling Changes
Using Isotope Analysis to Understand Phosphorus Cycling Changes
By Dr. Mario Tuthorn, Senior Product Marketing Specialist, Chromatography and Mass Spectrometry, Thermo Fisher Scientific 05.09.2024
Phosphorus is an essential nutrient for life, playing a role in various molecules and energy transfer in cells. It moves through the environment in biogeochemical cycles that transform and translocate phosphorus in soil, water, and living and dead organic material. The oxygen isotope composition (δ18O) of phosphate is a crucial tool for tracing biogeochemical phosphorus cycling.
However, the current methodologies for measuring this composition include tedious and time-consuming sample preparation, such as precipitating aqueous phosphate as silver phosphate (Ag3PO4). Besides challenging sample preparation, this method requires a large sample size and carries a risk of coprecipitation of other compounds like nitrate and dissolved organic compounds that can impact the quality of isotopic measurements.
A better way to analyze phosphate δ18O?
Is there a better, easier way to analyze phosphate δ18O? Lower your sample size with a new approach to phosphate isotope analysis.
A new study by Wang et al. (2024) presents a promising alternative. The research demonstrates the use of Thermo Scientific Orbitrap Electrospray Ionization Mass Spectrometry (ESI-Orbitrap-MS) for measuring the oxygen isotope composition of dissolved phosphate at nanomole level. This innovative method utilizes PO3 fragment ions to achieve comparable analytical precision to the conventional TC EA IRMS while using less than 1% of the sample amount.
Measuring phosphate δ18O using ESI-Orbitrap-MS methodology has been proposed by Neubauer et al. (2020), focusing on H2PO4– ions, however the results of the Wang et al. (2024) approach demonstrate that phosphate δ18O can be more reliably measured by the PO3− ions than by the H2PO4− ions. The authors highlight two main reasons:
  1. the H2PO4– (m/z = 97) can be interfered by the HSO4− (m/z = 97) that is often present at high concentrations in natural water samples, whereas P16O3− ions (m/z = 79) are not interfered with by 32S16O3− (m/z = 80) ions, and
  2. as PO3– fragment has no hydrogen, there is no need for high mass resolution mode (that is in H2PO4− ion measurements needed to separate H2P16O3 17O− and 2H1HP16O4−), resulting in higher analytical precision.
Wang et al. (2024) achieved a long-term accuracy of 0.8‰ (1 standard deviation) for phosphate δ18O under a dual inlet, direct liquid-phase injection condition by using two-magnitude smaller sample sizes (∼25 nmol) compared to the conventional IRMS method.
How can you expand your isotope research with this new, innovative workflow? Thermo Scientific Orbitrap Exploris Isotope Solutions comprises a dedicated sample introduction concept, distinctive method setup and a data evaluation framework to convert isotopologue intensities to accurate isotope ratios. Download the infographic for at glance overview of benefits of using this Orbitrap-based technology approach for deriving intramolecular isotopic information in biogeochemical research and beyond.
Additional isotope analysis content
Blog: How to Optimize Your Natural Abundance Isotope Ratio Analysis – Watch the Video
Blog: Trace Origin of Food with Strontium Isotope Analysis Using MC-ICP-MS
Blog: Isotope Fingerprints Fun Facts: How to Discover Origin and Authenticity with Isotope Analysis
On LinkedIn? Visit our LinkedIn page #isotopes #orbitrap #phosphorus #irms #environmental
From Complexity to Clarity: Smarter PFAS Identification Workflows in Compound Discoverer Software
Per- and polyfluoroalkyl substances (PFAS) have become a maj...
Read More
Innovation in Biopharma analysis image
Scaling Up Biopharmaceutical Characterization with the Orbitrap Ascend BioPharma Tribrid Mass Spectrometer
Biopharmaceutical pipelines are expanding rapidly, with incr...
Read More
Advancements in Thorium Isotope Analysis: The Role of Neoma MS/MS MC-ICP-MS
Thorium (Th) isotope analysis is important across various sc...
Read More
Dr Kerstin Krätschmer
GC Orbitrap Guru Series: Dr. Kerstin Krätschmer
It’s been 10 years! Yes, in 2015 “gas chromatography...
Read More
Dr. Mario Tuthorn
Dr. Mario Tuthorn is Senior Product Marketing Specialist for the Chromatography and Mass Spectrometry division of Thermo Fisher Scientific.
Leveraging LC-MS for Accurate Metabolite Identification (MetID) in Drug Discovery & Development
Back to Basics – Protein Characterization in Research and Pharmaceutical Development