Thermo Fisher Scientific
Electron and cryo-electron microscopy stories and solutions about the advancement of cellular and structural biology
Accelerating ScienceLife in Atomic Resolution / Electron Microscopy 101 / Seeing with Electrons: The Anatomy of an Electron Microscope
Seeing with Electrons: The Anatomy of an Electron Microscope
Written by Alex Ilitchev | Published: 11.04.2020
Light microscope vs electron microscope
While transmission electron microscopes (TEMs) continue to become more accessible thanks to novel automation, easy-to-use software, and decreasing space requirements, they can still be quite intimidating for new users who have never worked with an electron microscope before. They may be surprised to learn, however, that these instruments are not too different from the familiar light microscope.
Light microscope description
To understand how, let’s begin by considering the light microscope, shown below. In a light, or optical, microscope, a light source is placed below the sample, which must be thin enough to allow some light to pass through. (If it isn’t, the object will appear dark, and no details can be discerned.) The light is focused onto the sample with a condenser lens, resulting in a focused point of light on the specimen’s surface. As the white light interacts with the sample it is modified by its molecules, which selectively absorb or reflect only certain kinds of light. This results in the color and contrast information that makes up the image. Lenses then modify this projected information into a magnified image that we perceive with our eyes.
Light microscope diagram
Diagram of a light microscope with key elements highlighted.
Electron microscope description
In an electron microscope, an electron beam takes the place of the light source. Due to the wave-particle duality of electrons in a vacuum, they can behave just like the photons that make up light. However, electrons travel as much smaller wavelengths than visible light, enabling them to reveal smaller details than what is possible with light. Instead of a glass condenser lens, electron microscopes use electromagnetic or electrostatic lenses. These produce magnetic and electric fields that guide and focus the electrons. The sample is still positioned over a focused point of the incident beam but must now be even thinner to account for the size of the electrons. (Typical TEM sample thicknesses are around ~0.1µm, depending on the material.) Just like the optical microscope, the electron beam interacts with the sample atoms, carrying away contrast and compositional information. As our eyes are incapable of perceiving electrons, it is no longer sufficient to have them serve as the detector. Instead, special electron imaging devices convert the electron signal into a greyscale image, where contrast corresponds to differences in sample thickness and density.
Light microscope vs transmission electron microscope
Side-by-side comparison of a TEM and an optical microscope, with analogous parts connected. Note that the light microscope has been inverted to more clearly parallel the TEM structure.
Summary of instrument comparison
Overall, while there are different principles controlling and guiding the incident beam for both instruments, the process of image generation is broadly the same. A beam interacts with the sample and passes through to the other side; the changes in beam energy correspond to different features in the final image. Hopefully this quick comparison has dispelled some of the initial intimidation that electron microscopes pose, empowering you to pursue the unparalleled information that electron microscopy provides.
Luckily, the latest generation of cryo-transmission electron microscopes (cryo-TEMs) includes more affordable, easier-to-use tools. In late 2020, we introduced the Thermo Scientific Tundra Cryo-TEM which includes several features that improve usability and empower anyone to use cryo-EM, even researchers who are new to the technique.
Learn more about the Tundra Cryo-TEM >>
Alex Ilitchev, PhD, is a Scientific Content Writer at Thermo Fisher Scientific.
 //
 To learn more about electron microscopy for Life Sciences, please visit our Learning Center.
Decoding tauopathies: how cutting-edge cryo-EM is unraveling tau protein and Alzheimer’s disease
Tauopathy and neurodegeneration Neurodegenerative diseases a... by Shrikant Harne / 04.22.2025
Read More
Virus microscopy unravels chikungunya virus structure
Global impacts of the chikungunya virus Chikungunya is a vir... by Rosa Pepitone / 02.28.2025
Read More
cryoEM-blog-virus pore
What is cryo-EM?
Cryo-EM: cryo electron microscopy Cryo-electron microscopy (... by Alex Ilitchev / 10.30.2024
Read More
Next gen Glacios Cryo TEM helps solve CDK-cyclin at 2.3 Angstroms
Glacios 2 Cryo-TEM and the Evolution of 200 kV Cryo-EM
Glacios 2 Cryo-TEM and the Evolution of 200 kV Cryo-EM Cryo-... by Alex Ilitchev / 07.11.2024
Read More
Comments
  1. Mabel Cornwell says
    April 16, 2024 at 2:01 pm
    Hello,
    I am writing to see if I can get permission to use your image titled ‘Side-by-side comparison of a TEM and an optical microscope, with analogous parts connected. Note that the light microscope has been inverted to more clearly parallel the TEM structure’ for use in my PhD thesis.
    Thanks
    Reply
    • Alex Ilitchev says
      April 16, 2024 at 6:21 pm
      Hi Mabel,
      You may use the image, however, please include the following text somewhere near the image:
      “Illustration used courtesy of Thermo Fisher Scientific; copying prohibited.”
      Thank you and good luck with your thesis,
      Alex
      Reply
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Breakthrough Advances in Atomic Resolution Cryo-EM
Cryo-EM Accelerates Path to Understanding Diseases