Thermo Fisher Scientific
Molecular testing content for clinical lab professionals and clinicians
  • Categories
    • Science and education
    • Product news
    • Case studies
    • Perspectives
    • Events
    • News
Introducing UgenTec’s FastFinder Assay Plugins
Clinical Conversation blog - home icon Product news 3 mins. Sharing ▼ Details ▼
  • Linked In sharing icon
  • Facebook sharing icon
  • Twitter sharing icon
  • Email sharing icon
12 Sep 2022 || By Peter Friebe Shares: 1 Versions of this article Original article. Tags COVID-19, Influenza
UgenTec’s FastFinder Assay Plugin, for use with the TaqPath™ COVID 19 CE IVD RT PCR Kit, and TaqPath™ COVID-19, Flu A/B, RSV Combo Kit, is now available as a CE-marked device.
Applied Biosystems™ TaqPath™ COVID‑19 CE‑IVD RT‑PCR Kit
The TaqPath COVID‑19 CE‑IVD RT‑PCR Kit enables clinical and public health laboratories to quickly diagnose COVID-19, by the qualitative detection of nucleic acid from SARS-CoV-2 in upper respiratory specimens. This test uses a multi-target design which may be useful for identifying emerging variants.
Applied Biosystems™ TaqPath™ COVID-19, Flu A/B, RSV Combo Kit
The CE-IVD-marked, TaqPath COVID-19, Flu A/B, RSV Combo Kit detects and differentiates between SARS-CoV-2, influenza A/B, and RSV in a single sample. This test enables laboratories to expand their respiratory sample testing menu, ensuring proper patient care with timely and accurate diagnoses.
UgenTec’s FastFinder plugin is easy to use in conjunction with the TaqPath kits. Once the specimens are received by a lab, RNA is purified, reverse-transcribed into cDNA, and amplified on a real-time PCR instrument using the TaqPath kit and corresponding workflow. Raw PCR data are then transferred to the FastFinder software (rather than the Applied Biosystems Interpretive Software), which analyzes the data.
UgenTec’s FastFinder Analysis platform successfully hosts, processes, and displays the results of assay-specific plugins. FastFinder uses an assay-specific algorithm and decision mechanism to convert PCR raw data into test results. Cq values of each PCR curve are determined using a static model trained by machine learning technology, with cut-off Cq values built into the algorithm which are based off each TaqPath assay’s IFU. The software flags samples with suspicious amplification (such as a weak-positive sample with a strong-positive neighbor, or a cluster of positive samples) for manual review, and authorization is blocked until a trained professional has confirmed or rejected the suggested sample status.
To provide additional safeguards, plugins are self-contained and encrypted, and all assay-specific information (device type, assay type, used channels and targets, algorithm, and decision tree) is fixed within the assay plugin. The assay plugin offers full sample traceability across workflow steps, and all changes in the FastFinder software are audit trailed to maintain full data integrity.  Upon final review and authorization, results may be exported as input for an external software system such as LIMS.
A detailed overview of the performance validation performed by UgenTec can be found in the following white papers:
  • Performance Validation of UgenTec’s CE-IVD FastFinder Assay Plugin for the Applied Biosystems™ TaqPath™ COVID-19 CE-IVD RT-PCR Kit
  • Performance Validation of UgenTec’s CE-IVD FastFinder Assay Plugin for the Applied Biosystems™ TaqPath™ COVID-19, Flu A/B, RSV Combo Kit
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Home > Product news > Introducing UgenTec’s FastFinder Assay Plugins
Talk to us