Thermo Fisher Scientific
Molecular and antibody testing content for clinical lab professionals and clinicians
  • Categories
    • Science and education
    • Product news
    • Case studies
    • Perspectives
    • Events
    • News
Thermo Fisher’s COVID-19 tests: designed with virus mutations in mind
Clinical Conversation blog - home icon Science and education 4 mins. Sharing ▼ Details ▼
  • Linked In sharing icon
  • Facebook sharing icon
  • Twitter sharing icon
  • Email sharing icon
24 Dec 2020 || By Manoj Gandhi, M.D., Ph.D. Shares: 0 Versions of this article Original article. Tags COVID-19, Sars-cov-2, SGTF, Variants
This blog represents the most up-to-date information about the B.1.1.7 SARS-CoV-2 variant as of February 1, 2021.
Updated December 30, 2020: While the B.1.1.7 variant has become prevalent in parts of the United Kingdom, it is also beginning to emerge in other countries, including the United States1.As a leading supplier of COVID-19 testing solutions, Thermo Fisher Scientific is working with the US FDA to help with surveillance efforts for the new variant.
This post was written by Dr. Manoj Gandhi, MD, PhD, Sr. Medical Director at Thermo Fisher Scientific
A new SARS-CoV-2 variant has been identified
Reports of a new UK coronavirus variant, named by Public Health England as VOC202012/01 (the first “Variant of Concern” in December 2020) and also referred to as B.1.1.7 and 501Y.V1, have raised serious concerns as the world tries to understand what it might mean for transmission rates, disease severity, and more. Simultaneously, scientists are trying to understand how the many mutations might affect current SARS-CoV-2 diagnostic tests.
Mutations are the norm for viruses. In fact, high mutation rates in response to selective pressure is especially common in RNA viruses, and throughout the COVID-19 pandemic, SARS-CoV-2 has been mutating at a rate of about one to two mutations per month. What makes this new variant different is that it seems to have acquired mutations much more quickly than scientists expect resulting in a reported 70% increase in transmissibility.2 While B.1.1.7 shows increased transmissibility, preliminary evidence suggested there was no indication it was associated with increased disease severity.1 However, more recent studies do indicate that there is a realistic possibility of increased risk of death in patients infected with B.1.1.7.3
How does this new variant affect Thermo Fisher Scientific’s portfolio of COVID-19 diagnostics?
Knowing how common it is for viruses to mutate, we designed our COVID-19 molecular diagnostic tests in a multiplex manner; our assays target multiple areas of the SARS-CoV-2 genome. This built-in redundancy helps provide confidence that our tests can identify COVID-19 positive patients even when mutations in the genome arise.
Of the multiple mutations in this variant, 8 occur in the S gene that encodes for the spike (S) protein, while the others occur in other regions of the genome, such as in the orf1-ab, orf8, and nucleocapsid (N) genes. Only 1 mutation is important to be aware of in relation to the Applied Biosystems™ TaqPath™ COVID-19 assay—the 69-70del of the S gene.
Due to Thermo Fisher’s forward thinking 3-gene target assay design and the way that our software interprets results, if a sample infected with a variant that has the 69-70del mutation is tested using our TaqPath COVID-19 assay, it will result in an S gene “drop out.” This drop out does not mean a result is negative, only that the S gene could not be detected. Because of the multi-target design of our assays, to date, the 69-70del S gene mutation has not been found to impact test results obtained using the TaqPath COVID-19 tests, and the probability of a mutation in the S gene impacting the accuracy of our test results is low. Thermo Fisher’s team of scientists have been monitoring the variant situation closely to ensure that our assay continues to provide reliable results.
What is the value of using multi-target, real-time PCR diagnostics for COVID-19 testing?
The European CDC recommends that multi-target RT-PCR assays that include an S gene target affected by the deletions (and therefore will result in an S gene “drop out”) can be used as a signal for the presence of the 69-70del mutation for further investigation, especially if sequencing capacity is limited.2 Multi-target RT-PCR tests that utilize S gene regions impacted by the 69-70del mutation are quicker and cheaper than sequencing the entire virus and can help keep track of these mutant strains.4 If available, the European CDC recommends confirmation of the mutation using sequencing. 2
How to learn more
Thermo Fisher Scientific committed to providing our global customers with gold-standard molecular diagnostic technology. We regularly monitor post-market reports and public database updates to provide quality and accurate products.
If you’d like more details about the impact of the 69-70del S gene mutation observed in the B.1.1.7 variant on our TaqPath COVID-19 tests, please download our detailed FAQ document for the products currently available in your country:
  • FAQ for our CE-IVD marked tests
  • FAQ for our Emergency Use Authorized (EUA) test
If you have additional questions or would like to discuss your specific situation, please contact our technical support team at www.thermofisher.com/contactus.
References
  1. https://www.washingtonpost.com/health/coronavirus-variant-colorado-us/2020/12/29/8e6379fc-4a01-11eb-a9f4-0e668b9772ba_story.html
  2. https://www.ecdc.europa.eu/en/publications-data/threat-assessment-brief-rapid-increase-sars-cov-2-variant-united-kingdom
  3. https://assets.publishing.service.gov.uk/government/uploads/system/uploads/attachment_data/file/955239/NERVTAG_paper_on_variant_of_concern__VOC__B.1.1.7.pdf
  4. https://www.sciencemag.org/news/2020/12/mutant-coronavirus-united-kingdom-sets-alarms-its-importance-remains-unclear
Intended use and regulatory statements of products vary. For specific intended use and regulatory statements please refer to the Instructions for Use (IFU). Product availability by country varies. For product availability in your country, please check the TaqPath COVID-19 Multiplex Diagnostic Solution webpage.
Privacy StatementTerms & ConditionsLocationsSitemap
© 2026 Thermo Fisher Scientific. All Rights Reserved.
Home > Science and education > Thermo Fisher’s COVID-19 tests: designed with virus mutations in mind
Talk to us