Thermo Fisher Scientific
Molecular and antibody testing content for clinical lab professionals and clinicians
  • Categories
    • Science and education
    • Product news
    • Case studies
    • Perspectives
    • Events
    • News
Unraveling Solid Tumor Genetics with Whole-genome Copy Number Analysis
Clinical Conversation blog - home icon Science and education 3 mins. Sharing ▼ Details ▼
  • Linked In sharing icon
  • Facebook sharing icon
  • Twitter sharing icon
  • Email sharing icon
8 Jun 2024 || By Clinical Conversations Staff Shares: 4 Versions of this article Original article. Tags Oncology, OncoScan
Throughout the human genome, certain sections of DNA are repeated, and the number of these repeats varies between individuals. This molecular phenomenon, known as copy number variation (CNV), correlates with alterations of gene expression and is thought to play a significant role in the development and progression of cancer and other diseases. [1,2]
Researchers are now beginning to use copy number analysis to identify relevant CNVs, ones that could reveal new biomarkers and therapeutic targets for cancer treatments in the future.
A recent webinar provides important insights into how CNVs in solid tumor samples can be mapped, and it highlights the use of Applied Biosystems™ OncoScan™ CNV assays for research that involves profiling copy number alterations across the genome.
Watch the webinar
In the first half of the webinar, Joanna Przybyl, PhD, Senior Research Scientist at Stanford University, presents a strategy for profiling DNA from leiomyosarcoma (LMS) tumors. LMS is a rare and aggressive type of cancer that forms in smooth, involuntary muscles of the body including in the stomach, bladder, uterus, and blood vessels. As Dr. Przybyl explains, LMS is difficult to treat, in large part because of its characteristic high genomic instability, including intermediate levels of both DNA copy number alterations and somatic point mutations.
“The most important challenges in the treatment of patients with leiomyosarcoma are related to the lack of therapeutic targets in these tumors and also inadequate method for evaluation of
response to treatment,” she says.
Dr. Przybyl stresses that new therapeutic tools are needed. In addition to addressing studies from DNA extracted from FFPE samples, she describes research based on profiling molecular alterations in LMS tumor derived DNA in circulation, which may have potential as a non-invasive assessment of response to treatment in LMS patients. This approach combines detection of point mutations by deep targeted sequencing using a custom capture panel together with detection of copy number alterations through shallow whole genome sequencing.
Dr. Przybyl’s presentation includes details about how diploid recentering was used for correction of genomic profiles in tumors with high complexity and why genomic profiles characterized by single nucleotide polymorphism (SNP) array in tumor specimens may serve as a reference for detection of tumor-derived DNA copy number alterations in plasma.
In the second half of the webinar, Ravindra Kolhe, MD, PhD, Associate Professor, Pathology, Associate Director, Residency Program, Medical Director, Cytogenetics Laboratory at Augusta University, discusses the utility of whole genome single nucleotide polymorphism microarray (SNPM) in the evaluation of histologic mimics of melanoma.
Dr. Kolhe begins his presentation by explaining how factors such as the limitations of FISH testing and the prevalence of ambiguous histologic mimics can lead to both the under- and over- identification of melanoma. As a result, melanoma is particularly well-suited for research involving new tumor profiling tools, he says.
Dr. Kolhe walks through the SNPM workflow in detail and then goes on to discuss how these methods are being used to investigate other cancer types, including low-grade adult brain tumors and atypical lipomatous tumors.
“Whole genome SNP array can definitely make a substantial addition to the histologic evaluation of these small biopsies, especially differentiating between lipoma or liposarcoma,” he concludes.
To learn more about the importance of copy number analysis in LMS and melanoma, watch the webinar titled Exploring the genetic landscape of solid tumors using whole-genome copy number analysis for research. The methods described in the presentations are being developed to improve tumor profiling and identify new biomarkers.
To learn more about OncoScan CNV assays, please click here.
For Research Use Only
References:
  1. https://bmcmedgenet.biomedcentral.com/articles/10.1186/s12881-019-0909-5
  2. https://www.ncbi.nlm.nih.gov/pmc/articles/PMC3234280/
Privacy StatementTerms & ConditionsLocationsSitemap
© 2026 Thermo Fisher Scientific. All Rights Reserved.
Home > Science and education > Unraveling Solid Tumor Genetics with Whole-genome Copy Number Analysis
Talk to us