IAFP´s European Symposium, takes place at the La Cité des Congrès de Nantes, France this year (24th-26th April 2019). The IAFP’s European Symposium on Food Safety focuses on shaping the future of food safety by providing a forum for the exchange of ideas with colleagues from across Europe working in the industry, government, and academia. The event is an excellent forum to gain knowledge about the latest developments and techniques in food science and safety.
You will find Thermo Fisher Scientific exhibiting at Stand #19 where we will be demonstrating some of our most recent technology and solutions including a complete next generation sequencing (NGS) system for untargeted species screening of foods and ingredients; a powerful tool helping food manufacturers to combat food fraud.
Take a look at how to make sample handling and enrichment quick and easy with our Thermo Scientific™ Bagged Media solutions and how you can rapidly and effectively identify the most common foodborne pathogens with our Thermo Scientific™ SureTect™ Real-Time PCR System, now with workflows validated with the latest Applied Biosystems™ instrumentation and Thermo Scientific™ RapidFinder™ Analysis Software. Heating steps for the SureTect Assay sample preparation process can now be fully automated using the Applied Biosystems™ SimpliAmp™ Thermal Cycler. The SimpliAmp touchscreen interface makes it easy to set-up and save the parameters required for SureTect Assays and any other applications. The QuantStudio 5 Food Safety Instrument is a high-performance benchtop platform giving users greater control of data, combining out-of-the box installation, ease-of-use and system connectivity with powerful options to enable maximum control. The 6-channel, 96-well, cloud-enabled open platform is suitable for running the full range of PCR solutions for food safety, authenticity, and quality testing available from Thermo Fisher. Combining the QuantStudio 5 Food Safety Instrument with user-friendly RapidFinder Analysis software, makes SureTect Assay set-up, running and automatic result interpretation simple, and helps avoid errors and subjective result determination.
You’ll have the opportunity to find out more about the latest validation certifications and approvals for the SureTect pathogen detection assays and other Thermo Scientific workflows by visiting our stand and viewing our poster presentations:
P1-03: AOAC-RI PTM and NF Validation of the Thermo Scientific SureTect Salmonella Species PCR Assay Using the Quantstudio 5 PCR instrument
P1-04: NF Validation of the Thermo Scientific SureTect Cronobacter Species PCR Assay Using the Applied Biosystems Quantstudio 5 PCR Instrument
P1-05: NF Validation of the Thermo Scientific Listeria Precis Enumeration Method in Accordance with EN ISO 16140-2:2016
P1-06: NF Validation of the Thermo Scientific Listeria Precis Detection Method in Accordance with EN ISO 16140-2:2016
P1-07: Microval Validation of the Thermo Scientific Brilliance Staph 24 Enumeration Method in Accordance with ISO 16140-2:2016
P2-28: Microval Validation of the Thermo Scientific Brilliance CampyCount Enumeration Method in Comparison to EN ISO 10272-2:2017 in Accordance with ISO 16140-2:2016
Our team will be on hand to answer questions and provide updates on our newest solutions and more. We look forward to seeing everyone at IAFP this year.
Leave a Reply