How Does SEM EDS Support Technical Cleanliness Analysis in Automotive Parts Manufacturing?
Key takeaways
  • Technical cleanliness is a critical factor in product reliability, especially in high-precision industries such as automotive parts manufacturing; future cleanliness standards will increasingly require sub-micrometer detection
  • Compared to optical microscopy, scanning electron microscopy (SEM) offers superior resolution and material contrast; the addition of energy-dispersive X-ray spectroscopy (EDS) adds elemental characterization for improved particle classification and root cause identification
  • By integrating imaging and chemical analysis into a single workflow, SEM-EDS with the Thermo Scientific Axia ChemiSEM System enables more accurate contamination control and supports higher manufacturing quality standards
Why is technical cleanliness critical for automotive manufacturing?
Technical cleanliness in parts manufacturing is quantified by the presence, size, and composition of particulate contamination on the manufactured components. These particles can originate from the manufacturing process itself, handling and/or the assembly environment, as well as residual materials from machining or coating.
Technical cleanliness has become a critical quality parameter in modern manufacturing, especially in automotive, aerospace, and precision engineering. Even small particles can disrupt system performance, and as component tolerances tighten and system complexity increases, even micrometer-scale contamination can lead to premature wear, corrosion, or catastrophic failure. For example, metallic debris can cause abrasion in moving parts, hard particles such as Al₂O₃ can induce wear, and non-metallic contaminants can affect sealing or electrical behavior.
Standards such as ISO 16232 and VDA 19.1 define how particles should be collected, measured, and reported to ensure consistent quality control. This blog explores how scanning electron microscopy (SEM), combined with energy-dispersive X-ray spectroscopy (EDS), enhances particle analysis workflows to enable more reliable and actionable assessment of technical cleanliness.
Technical cleanliness from particle detection to classification
When it comes to the technical cleanliness of manufactured parts, contaminant particle size alone does not always tell the full story. In this application note, we compare optical microscopy and SEM-EDS workflows, showing how chemical identification enhances contamination analysis under ISO 16232 and VDA 19.1 standards.
Discover how automated SEM-EDS enables detection, measurement, and classification of particles in a single workflow, improving both speed and accuracy.
Read the “Electron microscopy in technical cleanliness analysis” application note >
How are particle contaminants collected and analyzed?
A typical technical cleanliness workflow begins with the extraction of particles from component surfaces, which can be done through a number of common methods, including particle measurement cards (PMCs), particle traps deployed in the production environment, and washing of components followed by filtering of the cleaning solution. Collected particles are deposited on a filter, where they can be analyzed with a variety of microscopy techniques. The filter is scanned field by field, particles are detected based on contrast, and their size and shape are measured.
Diagram showing particle analysis for technical cleanliness, including filter scanning, particle detection, and size measurement steps.
Overview of the particle analysis process used to assess technical cleanliness, including sample scanning, particle detection, and measurement.
Optical microscopy: applications and limitations for technical cleanliness
Optical microscopy is widely used for the assessment of technical cleanliness due to its speed and simplicity, and it can scan an entire filter in approximately 5 to 10 minutes. However, it has several limitations:
  • Optical detection depends on color contrast between particles and the background
  • Transparent or light-colored particles, such as glass and TiO₂, are difficult to detect visually
  • Limited resolution makes accurate measurement of small particles challenging
Additionally, optical microscopy does not provide compositional information, which can be critical for identifying the specific source and risk level of contaminants.
How does scanning electron microscopy improve particle contaminant detection?
Scanning electron microscopy uses backscattered electrons (BSE) to generate image contrast rather than visible light; BSE contrast is based on the molecular weight of the sample surface atoms. As a result, SEM can be used to detect particles regardless of their color, offering clearer differentiation of materials with varying atomic compositions. This can even be used to visualize particles that are otherwise invisible under optical microscopes, such as TiO₂ and glass. With SEM, these can be easily differentiated from a carbon-based filter background, since the heavier elements appear brighter in the BSE image.
SEM image showing the difference in contrast between aluminum oxide and pure aluminum particles on a carbon background.
SEM image showing the difference in contrast between aluminum oxide and pure aluminum particles on a carbon background.
How does scanning electron microscopy improve particle size measurement?
Standards such as ISO 16232 require the detection of particles down to 5 µm, and there is increasing demand for sub-micrometer analysis as well. Accurate measurement depends on pixel resolution. Optical microscopes often lack sufficient resolution for small particles, whereas SEM is capable of attaining nanometer-scale pixel resolution along with precise shape and size characterization. This ultimately offers more reliable detection of sub-micrometer particles, which is particularly important in high-precision applications such as automotive technical cleanliness, where even very small contaminants can cause failure.
What is the benefit of combined SEM EDS analysis?
A key advantage of SEM is that it can leverage energy-dispersive X-ray spectroscopy (EDS) as part of the electron imaging process. EDS adds the ability to obtain the elemental composition of each particle, allowing it to be classified not only by size and shape, but also by its chemical identity. With EDS, you can more easily identify particle origins, differentiate between similar-sized particles, and classify critical, harmful materials such as alumina (Al₂O₃) or silicon carbide (SiC). For example, two particles that appear to have the same size and SEM contrast can have vastly different impacts on the technical cleanliness of an automotive part, depending on whether they are soft polymer debris or hard ceramic contaminants.
Can scanning electron microscopy match the speed of optical microscopy?
Historically, SEM analysis has been slower than optical imaging due to the smaller fields of view obtained with EM, and the additional time needed for EDS steps. The Thermo Scientific Axia ChemiSEM System, combined with Thermo Scientific Perception Software, address these limitations by automating the scanning of whole filters while also integrating SEM and EDS analysis into a single run. Analysis of up to four filters can be done without user intervention.
Typically, the Axia ChemiSEM System can analyze a complete filter in approximately 20 minutes, and EDS acquisition adds only 0.1 to 1.0 seconds per particle. This significantly narrows the gap between SEM-EDS analysis and optical microscopy, while also generating far more detailed data.
How does automation improve the efficiency of technical cleanliness analysis?
Automated SEM workflows eliminate the need for manual particle selection, sample transfer between instruments, and lengthy correlation between optical and SEM datasets. Following data acquisition, particles can be filtered based on size, shape, or composition, and selected subsets (such as the largest or most critical particles) can be analyzed immediately. Reporting tools generate results aligned with ISO 16232 and VDA 19.1 standards, simplifying compliance and decision-making.
What does the future of technical cleanliness look like?
As manufacturing tolerances continue to shrink, cleanliness requirements are becoming more stringent. In some applications, particles smaller than 1 µm can impact performance and reliability. SEM-EDS analysis is well positioned to meet these demands due to its high-resolution imaging capabilities, integrated compositional analysis, and scalability with automated workflows. This makes scanning electron microscopes an essential tool for next-generation technical cleanliness analysis in advanced manufacturing environments.
Explore more contamination control and materials analysis solutions from Thermo Fisher Scientific
Technical cleanliness is only one part of a broader challenge in automotive manufacturing: ensuring materials and processes meet increasingly strict performance requirements.
Explore our “Advanced analysis in automotive manufacturing” eBook to learn how advanced analytical techniques, enabled by Thermo Scientific technology, support contamination control, defect analysis, and quality assurance across the full production cycle.
Download the eBook
Visit our technical cleanliness learning center for even more information and resources >
Alice Scarpellini
Written by:
Alice Scarpellini
Applications Development Scientist, Thermo Fisher Scientific
Alice Scarpellini is an Applications Development Scientist with more than 15 years of experience in advanced electron microscopy and microanalysis. She brings deep expertise in scanning electron microscopy, scanning transmission electron microscopy, energy-dispersive spectroscopy and electron backscatter diffraction and focuses on helping customers understand the scientific impact of these techniques.
Read more Scarpellini, Alice