Indonesia has no nuclear weapons or nuclear power plants, so how did Caesium 137, a radioactive isotope, enter Indonesia and contaminate a batch of shrimp, which ended up in the United States, possibly through scrap metal.
The Issue: Radiation Contamination of a Food Supply
Last Fall, it was reported by Reuters*, that Indonesia’s special task force on radioactive contamination discovered Caesium 137 at 22 production facilities in an industrial zone near Jakarta. The contamination was initially detected in shrimp shipped to the U.S. by PT Bahari Makmur Sejahtera (BMS). Following this, extensive radiation scans were conducted at the Modern Cikande Industrial Estate. BMS has since decontaminated its facility, but the names of the other 21 affected facilities have not been disclosed. The government plans to tighten restrictions on scrap metal imports, and a scrap metal factory, PT PMT, has been designated as an isolation facility for contaminated goods.
How Radiation Enters the Scrap Metal Stream
Medical diagnostic and monitoring equipment use radiological sources. Industries that make certain batteries, non-stick cookware, wrinkle-resistant fabrics, reflective signs, paper products, and many other products utilize radioactive substances and equipment that contains radioactive sources. Academic and scientific institutions use nuclear materials in their work. There is a wide range of industries and a wider variety of applications that utilize these sources.
But as in most industries, there is waste that needs to be disposed of, and if these sources are disposed of improperly, they can get mixed up in the waste stream and contaminate landfills and the soil. Even worse if they are in the recycling heap they could end up at metal recycling facilities, where they contaminate the materials they touch, then get melted all together and contaminate the whole batch – while contaminating the melting pot itself and all the equipment used to melt down the materials. And that’s after they have contaminated shredding and sorting equipment and conveyor systems, and before contaminating the vehicles used to transport them. Worst of all, they could jeopardize the health of the workers involved in all those steps.
The USA Environmental Protection Agency provides information on radioactive material in scrap metal, including what you can do and where to learn more, here: https://www.epa.gov/radtown/radioactive-material-scrap-metal.
Using Radiation Detectors and Dosimeters to Help Detect Contaminated Scrap Metal
To protect metal recycling facilities from orphan sources and radiation contamination, it’s crucial to prevent radioactive materials from entering the facility. There are several types of radiation detection and measurement instruments and technologies available. Automatic Scrap Monitoring Systems can detect radiation in vehicles and processes within the facility, providing accurate and reliable detection with a low false alarm rate. These systems monitor trucks, vehicles, railcars, conveyors, platform scales, and dust collection equipment to reject radioactive materials before they pose a threat to people or the environment. There are at least 10 separate places in a recycling yard where radiation detection and monitoring equipment can be effectively used.
To protect workers, radiation detection devices and dosimeters can be utilized.
- Radiation Detection Devices: These are instruments used to detect and measure the presence of radiation. They can identify radioactive materials and measure radiation levels in an environment. Examples include Geiger counters, scintillation detectors, and ionization chambers.
- Dosimeters: These are devices used to measure an individual’s exposure to radiation over time. They are typically worn by personnel working in environments where radiation is present to monitor cumulative exposure. Examples include film badges, thermoluminescent dosimeters (TLDs), and electronic personal dosimeters.
Handheld radiation detection devices provide real time detection of gamma radiation with accurate dose rate measurements to verify the radioactive find and assess whether radioactivity is of natural or artificial (manufactured) origin. Portable devices with high sensitivity neutron response and alarm threshold can be worn to monitor gamma sensitivity and energy compensated dose rate measurement.
We have identified eight key places to consider when looking to protect scrap metal recycling facilities and the public from radiation threats. See the infographic here.
Conclusion
The discovery of Caesium 137 contamination in Indonesia underscores the critical need for stringent monitoring and regulation of scrap metal imports to prevent radioactive materials from entering the supply chain. Implementing advanced radiation detection systems and dosimeters in recycling facilities can effectively identify and mitigate contamination risks, safeguarding public health and the environment. This incident highlights the importance of global vigilance and cooperation in managing radioactive materials, helping to ensure that industries adhere to safety protocols to prevent similar occurrences in the future.
References and Additional Resources
- Infographic: 8 Reasons (and Places) Metal Recyclers Need Radiation Detection
- Brochure: Industrial Radiation Protection for Your Personnel, Facility and Business
- Radiation detection and measurement instruments and technologies.
- Radiation monitoring and detection technologies for scrap metal recycling companies.
- *https://www.reuters.com/sustainability/boards-policy-regulation/indonesia-says-22-plants-industrial-zone-near-jakarta-contaminated-by-caesium-2025-10-08/
- *https://www.reuters.com/world/asia-pacific/indonesia-says-it-is-touch-with-nuclear-watchdog-united-states-radioactive-2025-09-30/
FREQUENTLY ASKED QUESTIONS
1. How does radioactive material end up in scrap metal?
Radioactive sources from medical devices, industrial gauges, research equipment, or improperly disposed materials can enter the scrap stream as “orphan sources.” When mixed with recyclable metal, they can contaminate loads, equipment, and facilities.
2. What are the most common radioactive isotopes found in scrap?
Common isotopes include Cesium-137, Cobalt-60, Iridium-192, Americium-241, and Radium-226. These are typically used in medical therapy, industrial radiography, density gauges, and other industrial applications.
3. How can we detect radiation before it contaminates our facility?
Recyclers use radiation portal monitors, handheld detectors, and personal dosimeters. Portal systems scan incoming trucks and railcars, while handheld devices verify and localize radioactive sources.
4. What happens if radioactive scrap is melted?
If a radioactive source enters a furnace, it can contaminate the entire melt, slag, baghouse dust, furnace lining, and surrounding equipment. Cleanup is expensive and may shut down operations for extended periods.
5. What are the regulatory requirements for radiation in scrap metal?
Regulations vary by country, but recyclers are typically required to report radioactive findings to national or regional authorities and follow proper isolation and disposal procedures. Environmental and worker safety regulations also apply.
6. How can we protect workers from radiation exposure?
Facilities implement layered protection: portal monitoring at entry points, handheld verification tools, clear response procedures, training programs, and personal dosimeters to monitor cumulative exposure.
7. What should we do if radiation is detected in a load?
Immediately isolate the material, secure the area, prevent further handling, notify management and authorities, and use trained personnel with proper detection equipment to assess and manage the source safely.
Leave a Reply