Seth Gold Tells All: National Pawnbroker of the Year Explains the Need to Drop the Acid
Seth Gold of American Jewelry and Loan  2013 NPA Pawnbroker of the Year and reality television personality  recently sat down with me and explained why XRF equipment is a must-have technology for pawnbrokers. Here’s what he had to say:
Drop the Acid
by Seth Gold, American Jewelry and Loan
Seth Gold
Seth Gold
My family and I own a jewelry store in Detroit and run a ton of transactions. Many of our customers come in to pawn or sell us their old gold jewelry. Much of the jewelry is out of fashion or broken so there’s no resale value. We send this scrap jewelry to a refiner to be melted down.  Prices are usually based on the weight of the gold content. Solid gold jewelry is 24 karats. Lesser jewelry has less gold content and usually contains other metals and hardening agents.  The less gold content in a piece of jewelry, the less money it will be worth to anyone who intends to melt it down.  So if I receive less money from the refiner, I will have to offer less money to the customer.
To stay in business, I need to make sure the cash offer is fair to both the customer and to me.  Before I hand over money to the customer, I need to know exactly how much gold is in that scrap jewelry.  If the customer is mistaken about the karat weight, or if karat marks are wrong and I don’t know the exact content, I could be burned.
So how do I figure out how much gold is in the jewelry?  Well, not with acid tests anymore.  Acid is archaic.  Before we found XRF technology, we used acids.  First you have to scratch the jewelry on a stone to get flakes or a streak of gold that can be tested.  You are literally rubbing off some of the gold, and you need to make a deep enough scratch to be sure the jewelry isn’t gold-plated.  Then you put a few drops of corrosive acid on the gold streak.  I’ll say that again.  It’s corrosive. It’s dangerous. You need to wear gloves.  If it touches your skin, it can burn. You’d better not have any kids or pets around because it’s a caustic chemical, which means you also need good ventilation in your shop.  And you’d better make sure the surface you’re using won’t be destroyed.  Like I said, acid testing is archaic. You’re destroying the jewelry; you’re burning your fingers, burning your rugs, and creating a mess.  You might even be opening yourself up to employee liability.
To top it off, acid testing isn’t always accurate.  Most acid kits contain materials to test 10k, 14k, 18k,and 22k gold. The acid rounds to the nearest testing solution; it can’t tell you if it’s 13K or 18.5K.You have to trust that your employees will know all this and interpret the results correctly. Also, certain types of stainless steel sometimes pass the stone test for 18 karat gold. These days, counterfeiting is getting harder to spot. I know many shop owners who were deceived by alloy chains that are fooling the acid test.
When you’re dealing in precious metals  which is very competitive  accurate analysis can make all the difference.  If you have a precise read of the content, you might be able to pay your customers a little bit more and get a bigger return.  When you use the right technology, like XRF analysis, you can base your judgment on actual readouts.  You’re not guessing on what the material is.  You’re not relying on the stamp, which could be forged.  You are basing your decision on actual content.
How many times has a customer come in with a piece of jewelry that has more than one metal in it?  If a piece of jewelry is white and yellow, are you able to determine the actual karat on each color?  Is it platinum?  Is it white gold?  Is it yellow gold? My XRF equipment has technology that can zoom in on a small spot so it can determine what’s platinum, what’s gold, and if there are any other metals in it. That could mean a larger loan, or a smaller loan.
The XRF analyzer that we use also has special patented technology that alerts me if the piece is gold plated. If copper, steel, tungsten or any other non-gold substrate makes up that piece of jewelry, I’ll know it.
And you can show your customer the actual test results.  We have our XRF analysis equipment right there on the showroom counter.  It takes just a few seconds, and the customer can watch the analyzer as it tests the jewelry right in front of them.  They can’t question us about going in the back and switching their jewelry, or giving them less money than it’s worth.  That makes them trust us, so they will keep coming back to us.
I understand that most pawnbrokers don’t like spending money, and this technology has a big upfront cost, although some analyzers can be leased.  I don’t like spending money needlessly either.  But this is a cost of doing business; it will reduce your risk and it will pay for itself. If I told you that you could spend $20,000 but in the end you would save $100,000, you would be crazy not to take advantage of that opportunity. The technology is here. Use it.
Seth will be appearing at the upcoming National Pawnbrokers Association show:
Pawn Expo
Booth #205 – check schedule for times
July 8-10, 2014
Las Vegas, Nevada USA
Jonathan Margalit
Written by:
Jonathan Margalit, PhD
Business Development Manager, Thermo Fisher Scientific
Jonathan Margalit drove business development initiatives for Thermo Scientific portable analytical instrumentation in the environmental, consumer goods, mining, and metals markets from 2011-2016.
Read more Margalit, Jonathan
Comments
  1. Rich says
    Yeah, and XRF only goes a few microns below the surface so not helpful in detecting anything that’s heavily plated.
    • nitonxrf says
      Rich–
      Thank you for your comment.
      All portable XRF analyzers operate on a low power x-ray tube. While this, indeed, limits the penetration depth of the x-rays to about 10-15 microns (in the case of gold), it has the great advantage of keeping the analyzer small, and thus portable, as well as minimizing the regulatory obligations on behalf of the user for using x-rays. Note that some of the commercial portable XRF analyzers are equipped with a sophisticated and patented gold-plating detection technology that provides an additional layer of confidence in the detection of gold on flat and non-flat jewelry.
      Jonathan Margalit, PhD
      Business Development Manager – Precious Metals & Mining
      Thermo Scientific Portable Analytical Instruments
    • nitonxrf says
      Rich–
      “While a few portable EDXRF suppliers claim to have gold plating
      detection technology, not all methods are equally effective. Most of
      these plating detection techniques just look for high Ni content and/or
      non-traditional or unexpected karat values. One supplier, however, also
      adds an additional and third level of sophistication in a patented solution as
      shown in this video:
      http://youtu.be/1Fu8uLBdgcE?list=PLI4Chh5REK_THtDnZzICQEL6EUN0_23vW.
      In the uncommon case of plating more than 10-15 µm in thickness, the only
      viable portable method for detection that I am aware of is ultrasonic – but
      this requires some training and interpretation, and is limited mostly to very
      flat pieces.”
      Here’s some additional information:
      http://www.niton.com/en/metal-and-alloy-analysis/applications/goldanalysis
    • nitonxrf says
      FROM Rich:
      How does it detect gold-plating? XRF is good, just so darn
      expensive from you guys.
      4:58 p.m., Wednesday Dec. 10
  2. scott nell says
    It is so amazing post and very helpful for us as a pawn broker and for the costumers. It speaks clearly about how to detect how much gold does a jewelry has by using XRF technology. This is something we would love to use. Thank you sharing it with us.
  3. Jonas Toscaño says
    It really interests me to read such articles, thanks for sharing. Blogs like yours really helps me learn more day by day.